Skip to Content
Merck
All Photos(2)

Key Documents

B82251

Sigma-Aldrich

Bromotrichloromethane

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrCCl3
CAS Number:
Molecular Weight:
198.27
Beilstein:
1732543
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

vapor density

6.85 (vs air)

Quality Level

vapor pressure

38.4 mmHg ( 25 °C)

Assay

99%

form

liquid

refractive index

n20/D 1.5065 (lit.)

bp

105 °C (lit.)

mp

−6 °C (lit.)

density

2.012 g/mL at 25 °C (lit.)

SMILES string

ClC(Cl)(Cl)Br

InChI

1S/CBrCl3/c2-1(3,4)5

InChI key

XNNQFQFUQLJSQT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Chain transfer agent for radical polymerization of methacrylates. Brominating reagent.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Journal of the Chemical Society. Chemical Communications, 747-747 (1994)
Macromolecules, 27, 5863-5863 (1994)
Tetrahedron, 50, 31-31 (1994)
C Jochmann et al.
Pharmacology & toxicology, 75(1), 7-16 (1994-07-01)
The role of glutathione (GSH) and protein thiols in the pathobiochemical process of CBrCl3 cytotoxicity was investigated in isolated hepatocytes. Administration of 0.5, 1.0 and 1.5 mmol/l CBrCl3 affected cellular viability as assessed by trypan blue exclusion, release of lactate
Inhibition of vitamin B12-dependent methionine biosynthesis by chloroform and carbon tetrachloride.
T A Alston
Biochemical pharmacology, 42(12), R25-R28 (1991-11-27)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service