Skip to Content
Merck
All Photos(1)

Key Documents

940127

Sigma-Aldrich

Cesium chloride

new

≥99.5 (titration), ≥99.9 trace metals basis

Synonym(s):

Caesium chloride, Cesium monochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CsCl
CAS Number:
Molecular Weight:
168.36
MDL number:
UNSPSC Code:
12352200
Assay:
≥99.5% (with AgNO3, titration)
≥99.9 trace metals basis
form:
powder or crystals

Quality Level

Assay

≥99.5% (with AgNO3, titration)
≥99.9 trace metals basis

form

powder or crystals

impurities

≤1500 ppm (trace metals analysis)

mp

200 °C
645 °C (lit.)

density

2.19 g/cm3

cation traces

K: ≤0.05%
Li: ≤50.0 ppm
Mg: ≤1.0 ppm
Na: ≤0.1%
Ni: ≤1.0 ppm
Pb: ≤1.0 ppm
Rb: ≤0.1%

application(s)

(Energy)

SMILES string

[Cl-].[Cs+]

InChI

1S/ClH.Cs/h1H;/q;+1/p-1

InChI key

AIYUHDOJVYHVIT-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Cesium chloride is a highly water soluble crystalline solid withhigh refractive index and dielectric properties. It is widely used in the field of solid oxide fuel cells and lighting devices.[1,2]

Application

Cesium chloride can be used: -As crystal seeds to enhance the coverage of the CH3NH3PbI3−xClx absorber layer in perovskite solar cells. It helps to boost the power conversion efficiency of solar cells.[1] -As a medium to synthesize N and P-codoped carbon nanospheres for application in fuel cells and zinc-air batteries.[3] -To fabricate silicon nanopillars for solar cells, batteries, light-emitting diodes, etc.[4] -To enhance the thermal stability of polyoxometalates(used as electrode materials for lithium-ion batteries).[2] -To synthesize the lead-free halide double perovskite Cs2AgInCl6 with excellent moisture, light, and heat resistance. This material shows great potential for various optoelectronic applications, including photovoltaics.[5]

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Metal-Free N and P-Codoped Carbon Nanosphere as Bifunctional Electrocatalyst for Rechargeable Zinc-Air Batteries
Ping Li, et al.
ChemElectroChem, 6, 393-397 (2019)
Effect of cesium chloride modification on the film morphology and UV-induced stability of planar perovskite solar cells
Wenzhe Li et, al.
Journal of Materials Chemistry, 4, 11688-11695 (2016)
Fabrication and reflection properties of silicon nanopillars by cesium chloride self-assembly and dry etching
Jing Liu et, al.
Applied Surface Science, 258, 8825-8830 (2012)
Composition design, optical gap and stability investigations of lead-free halide double perovskite Cs2AgInCl6
Jun Zhou, et al.
Journal of Materials Chemistry, 5, 15031-15037 (2017)
Study of Polyoxometalates as Electrode Materials for Lithium-Ion Batteries: Thermal Stability Paves the Way to Improved Cycle Stability
Georg-Maximilian Bosch, et al
ChemElectroChem, 8, 656-664 (2021)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service