8.17006
Polyethylene glycol
4000, powder, EMPROVE® EXPERT, Ph. Eur., NF
Pharma Manufacturing
Synonym(s):
Poly(ethylene glycol), Polyglycol, Polyethylene oxide, Polyoxy ethylene, PEG 4000, PEG
About This Item
Agency
NF
Ph. Eur.
Quality Level
vapor pressure
<0.01 hPa ( 20 °C)
product line
EMPROVE® EXPERT
form
powder
solid
autoignition temp.
>320 °C (DIN 51794)
mol wt
average Mn 4000
technique(s)
API processing | nano-milling: suitable
pH
5-7 (20 °C, 100 g/L in H2O)
mp
53-58 °C
transition temp
flash point 270 °C
density
1.2 g/cm3 at 20 °C
application(s)
liquid formulation
pharma/biopharma processes
pharmaceutical
semi-solid formulation
solid formulation
solubility enhancement
shipped in
ambient
storage temp.
15-25°C
SMILES string
C(CO)O
InChI
1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2
InChI key
LYCAIKOWRPUZTN-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.
Application
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
281.5 °F - closed cup
Flash Point(C)
138.6 °C - closed cup
What is the Emprove® Program?
The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:
Free of charge - Sign in to download
Free of charge - Sign in to download
Available by subscription or for a fee
Available by subscription or for a fee
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
3D printing can overcome challenges during formulation development, with a focus on enhancing API bioavailability in solid dispersions. Find out how in this technical article.
3D printing can overcome challenges during formulation development, with a focus on enhancing API bioavailability in solid dispersions. Find out how in this technical article.
3D printing can overcome challenges during formulation development, with a focus on enhancing API bioavailability in solid dispersions. Find out how in this technical article.
3D printing can overcome challenges during formulation development, with a focus on enhancing API bioavailability in solid dispersions. Find out how in this technical article.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service