Skip to Content
Merck
All Photos(7)

Key Documents

C4036

Sigma-Aldrich

Cesium chloride

for molecular biology, ≥98%

Synonym(s):

CsCl, cesium monochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CsCl
CAS Number:
Molecular Weight:
168.36
EC Number:
MDL number:
UNSPSC Code:
12352302
eCl@ss:
38050206
PubChem Substance ID:
NACRES:
NA.31
grade:
Molecular Biology
for molecular biology
form:
powder

grade

Molecular Biology
for molecular biology

Assay

≥98%

form

powder

mp

645 °C (lit.)

foreign activity

DNase, RNase, protease, none detected

SMILES string

[Cl-].[Cs+]

InChI

1S/ClH.Cs/h1H;/q;+1/p-1

InChI key

AIYUHDOJVYHVIT-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Cesium chloride has been used in the generation of discontinuous gradient for the purification of Cryptosporidium oocysts and as a component of acetamide medium for fungal selection.
Used for the preparation of electrically conducting glasses.Used to make solutions for the separation of RNA from DNA by density gradient centrifugation.

Biochem/physiol Actions

Cesium chloride regimen results in controlling cancer cell growth by targeting alkalinization of the neoplastic cells. It elicits potassium-blocking and is used animal cardiovascular research.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Photocatalytic inactivation of the waterborne protozoan parasite Cryptosporidium parvum using TiO2/H2O2 under simulated and natural solar conditions
Abeledo-Lameiro MJ, et al.
Catalysis Today, 280(9), 132-138 (2017)
Cesium chloride-induced torsades de pointes
Wiens M, et al.
The Canadian Journal of Cardiology, 25(9), e329-e329 (2009)
Fatal cesium chloride toxicity after alternative cancer treatment
Sessions, et al.
Journal of Alternative and Complementary Medicine (New York, N.Y.), 19(12), 973-975 (2013)
Agrobacterium-mediated transformation of the filamentous fungus Aspergillus awamori.
Michielse CB, et al.
Nature Protocols, 3(10), 1671-1671 (2008)
Takafumi Yamamoto et al.
Inorganic chemistry, 50(22), 11787-11794 (2011-10-25)
The rock salt (B1) structure of binary oxides or chalcogenides transforms to the CsCl (B2) structure under high pressure, with critical pressures P(s) depending on the cation to anion size ratio (R(c)/R(a)). We investigated structural changes of A(2)MO(3) (A =

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service