Skip to Content
Merck
All Photos(2)

Documents

934755

Sigma-Aldrich

NMC622

greener alternative

electrode sheet, aluminum substrate, size 5 in. × 10 in.

Synonym(s):

Lithium Nickel Manganese Cobalt Oxide, NMC 622 cathode

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
LiNi0.6Mn0.2Co0.2O2
Molecular Weight:
96.93
UNSPSC Code:
26111700
NACRES:
NA.21

material

aluminum substrate

Quality Level

grade

battery grade

description

3.75 V vs Li/Li+
Application: Battery Manufacturing

form

sheet

composition

Active material loading, ≥85%

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

size

16 μm , aluminum current collector
5 in. × 10 in.

thickness

60 μm , excluding current collector

particle size

10-13 μm (active material characteristic)

capacity

2.0 mAh/cm2±5 % (Areal capacity)

application(s)

battery manufacturing

greener alternative category

General description

NMC622, electrode sheet, aluminum substrate, is a ready-to-use cathode for lithium-ion battery research. NMC622 is a quaternary lithium metal oxide, LiNi0.6Mn0.2Co0.2O2, and is a state-of-the-art cathode material for lithium-ion batteries that offers high energy density and cycle lifetimes. The composition of our cathode film is 90% active material, 5% PVDF binder, 5% carbon black.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more information.

Application

The main application of our NMC622 electrode sheet is as a cathode for next-generation lithium-ion batteries (LIBs). The 622 refers to the unique composition of the active material in which nickel provides high energy density while the manganese and cobalt help to stabilize the spinel crystal structure. As a result, our cathode sheet achieves high capacity (>166 mAh/g gravimetric capacity, 2.0 mAh/cm2 areal capacity) and long cycle lifetimes, while offering a high nominal voltage of 3.75 V vs. Li/Li+. The recommended charge rate for our sheet is 1C and discharge rate up to 5C.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peiyu Hou et al.
Small (Weinheim an der Bergstrasse, Germany), 13(45), 1701802-1701802 (2017-10-05)
The urgent prerequisites of high energy-density and superior electrochemical properties have been the main inspiration for the advancement of cathode materials in lithium-ion batteries (LIBs) in the last two decades. Nickel-rich layered transition-metal oxides with large reversible capacity as well
Seong-Min Bak et al.
ACS applied materials & interfaces, 6(24), 22594-22601 (2014-11-25)
Thermal stability of charged LiNixMnyCozO2 (NMC, with x + y + z = 1, x:y:z = 4:3:3 (NMC433), 5:3:2 (NMC532), 6:2:2 (NMC622), and 8:1:1 (NMC811)) cathode materials is systematically studied using combined in situ time-resolved X-ray diffraction and mass spectroscopy

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service