Skip to Content
Merck
All Photos(2)

Key Documents

229520

Sigma-Aldrich

Cadmium nitrate tetrahydrate

99.997% trace metals basis

Synonym(s):

Nitric acid, cadmium salt tetrahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cd(NO3)2 · 4H2O
CAS Number:
Molecular Weight:
308.48
Beilstein:
8156730
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.997% trace metals basis
form:
solid

Quality Level

Assay

99.997% trace metals basis

form

solid

reaction suitability

reagent type: catalyst
core: cadmium

impurities

≤35.0 ppm Trace Metal Analysis

mp

59.5 °C (lit.)

SMILES string

[H]O[H].[H]O[H].[H]O[H].[H]O[H].[O-][N+](=O)O[Cd]O[N+]([O-])=O

InChI

1S/Cd.2NO3.4H2O/c;2*2-1(3)4;;;;/h;;;4*1H2/q+2;2*-1;;;;

InChI key

QOYRNHQSZSCVOW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Muta. 1B - Repr. 1B - STOT RE 1

Target Organs

Kidney,Bone

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ali Hossain Khan et al.
ACS nano, 14(4), 4206-4215 (2020-04-11)
Colloidal two-dimensional (2D) nanoplatelet heterostructures are particularly interesting as they combine strong confinement of excitons in 2D materials with a wide range of possible semiconductor junctions due to a template-free, solution-based growth. Here, we present the synthesis of a ternary
E Helene Larsson et al.
Journal of experimental botany, 53(368), 447-453 (2002-02-16)
In order to test the potential effect of prior exposure to different Cd concentrations on Cd uptake and accumulation, plants of Arabidopsis thaliana, including a phytochelatin-deficient mutant, cad1-3, and the wild type, were compared. For Cd uptake experiments, plants were
Xue-Mei Li et al.
Journal of inorganic biochemistry, 101(8), 1165-1171 (2007-06-23)
A novel hybridization indicator, bis(benzimidazole)cadmium(II) dinitrate (Cd(bzim)(2)(NO(3))(2)), was utilized to develop an electrochemical DNA biosensor for the detection of a short DNA sequence related to the hepatitis B virus (HBV). The sensor relies on the immobilization and hybridization of the
Francesco Crea et al.
Annali di chimica, 95(11-12), 767-778 (2006-01-10)
In order to continue the investigation on the formation of hetero-metal polynuclear hydrolytic species, in this paper we report some results (at I = 0.16 mol L(-1) in NaNO3, at t = 25 degrees C by potentiometry, ISE-H+, glass electrode)
M S Monteiro et al.
Ecotoxicology and environmental safety, 72(3), 811-818 (2008-10-28)
Laboratory and field studies have provided encouraging insights into the capacity of plants to act as biomonitors of environmental quality through the use of biomarkers. However, a better understanding of the overall process of Cd-induced senescence, describing the cascade of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service