Skip to Content
Merck
All Photos(1)

Key Documents

746169

Sigma-Aldrich

Difluoro(4-(1,1-dimethylethyl)-2-{1-[4-(1,1-dimethylethyl)-3,5-dimethyl-2H-pyrrol-2-ylidene-N]ethyl}-3,5-dimethyl-1H-pyrrol-2-ylidene-N]ethyl}-3,5-dimethyl-1H-pyrrolato-N)boron

98% (HPLC)

Synonym(s):

Boron,[4-(1,1-dimethylethyl)-2-[1-[4-(1,1-dimethylethyl)-3,5-dimethyl-2H-pyrrol-2-ylidene]ethyl]-3,5-dimethyl-1H-pyrrolato-N1,N2]difluoro-, PM 570, PM 596, PM 597, Pyrromethene 570, Pyrromethene 596, Pyrromethene 597

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H33BF2N2
CAS Number:
Molecular Weight:
374.32
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

98% (HPLC)

form

powder

mp

256-261 °C

λmax

526 nm in dichloromethane

SMILES string

CC(C1=C(C)C(C(C)(C)C)=C(C)N1B(F)2F)=C3[N]2=C(C)C(C(C)(C)C)=C3C

InChI

1S/C22H33BF2N2/c1-12-17(21(6,7)8)15(4)26-19(12)14(3)20-13(2)18(22(9,10)11)16(5)27(20)23(26,24)25/h1-11H3

InChI key

SEHGNHOGQDPQRC-UHFFFAOYSA-N

General description

Difluoro(4-(1,1-dimethylethyl)-2-{1-[4-(1,1-dimethylethyl)-3,5-dimethyl-2H-pyrrol-2-ylidene-N]ethyl}-3,5-dimethyl-1H-pyrrol-2-ylidene-N]ethyl}-3,5-dimethyl-1H-pyrrolato-N)boron (BODIPY) is a boron containing dipyrromethene dye that can be used as a laser dye which exhibits emission from green to red region of a spectrum. It has good electrochemical and spectroscopic properties and can be used for a variety of light sensing applications.

Application

BODIPY dye can be used as a laser dye and finds application in the bio-labeling of DNA, proteins and lipids. It may also be used in the fabrication of position sensitive devices(PSDs) such as machine tools and aligning systems. It has a lasing wavelength of <500nm and can potentially be utilized in the development of photo-initiator systems.
BODIPY laser dyes are used to generate fluorescent conjugates of proteins; nucleotides; oligonucleotides and dextrans; and to prepare fluorescent enzyme substrates; fatty acids; phospholipids; lipopolysaccharides; receptor ligands and polystyrene microspheres.
Also used for solid-state dye laser devices and organic solar cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Electrogenerated chemiluminescence 71. Photophysical, electrochemical, and electrogenerated chemiluminescent properties of selected dipyrromethene- BF2 dyes
Lai RY and Bard AJ
The Journal of Physical Chemistry B, 107(21), 5036-5042 (2003)
High-efficiency pyrromethene doped solid-state dye lasers
Hermes RE, et al.
Applied Physics Letters, 63(7), 877-879 (1993)
Markus Franke et al.
ACS applied materials & interfaces, 9(34), 29193-29202 (2017-08-08)
Electrically tunable microcavities are essential elements for tunable laser sources indispensable for modern telecommunication and spectroscopy. However, most device concepts suffer from extensive lithography or etching for membrane processing. Here, we present an electrically and continuously tunable, multi-half-wavelength microcavity with
Conformable large-area position-sensitive photodetectors based on luminescence-collecting silicone waveguides
Bartu P, et al.
Journal of Applied Physics, 107(12), 123101-123101 (2010)
Holographic polymer dispersed liquid crystal reflection gratings formed by visible light initiated thiol-ene photopolymerization
Natarajan LV, et al.
Polymer, 47(12), 4411-4420 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service