Skip to Content
Merck
All Photos(2)

Key Documents

790389

Sigma-Aldrich

Difluoro{2-[1-(3,5-dimethyl-2H-pyrrol-2-ylidene-N)ethyl]-3,5-dimethyl-1H-pyrrolato-N}boron

99% (HPLC)

Synonym(s):

1,3,5,7,8-Pentamethyl-4,4-difluorro-4-bora-3a,4a-diaza-s-indacene, 2-[1-(3,5-Dimethyl-2H-pyrrol-2-ylidene)ethyl]-3,,5-dimethyl-1H-pyrrole, boron complex, 4,4-Difluoro-1,3,5,7,8-pentamethyl-4-bora-3a,4a-diaza-s-indacene, BODIPY 493/503, PM 546, PMPBF2, Pyrromethene 546, [[2-[1-(3,5-Dimethyl-2H-pyrrol-2-ylidene)ethyl]-3,5-dimethyl-1H-pyrrolato-N1,N2]difluoro]boron

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H17BF2N2
CAS Number:
Molecular Weight:
262.11
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

99% (HPLC)

form

powder

mp

262-266 °C

fluorescence

λex 493 nm; λem 504 nm in methanol

SMILES string

CC(C1=C(C)C=C(C)N1B(F)2F)=C3[N]2=C(C)C=C3C

InChI

1S/C14H17BF2N2/c1-8-6-10(3)18-13(8)12(5)14-9(2)7-11(4)19(14)15(18,16)17/h6-7H,1-5H3

InChI key

DRJHPEGNOPSARR-UHFFFAOYSA-N

General description

Difluoro{2-(1-(3,5-dimethyl-2H-pyrrol-2-ylidene-N)ethyl)-3,5-dimethyl-1H-pyrrolato-N}boron is an organoboron composed of dipyrromethene with a boron in the center. It is a fluorescent dye with a high quantum yield and an ability to absorb small molecules with sharp peaks in the ultraviolet region.

Application

BODIPY can be infused in silicon oil to impart fluorescence which can be used in light microscopy for cell sheets. It can be used in labelling of mouse pre-adipocyte 3t3-L1 cells. It may also be used in combination with bipyridine copper based complex which can be used in the sensing of nitrogen oxide(NOx) in air and living cells.
BODIPY laser dyes are used to generate fluorescent conjugates of proteins, nucleotides, oligonucleotides and dextrans, and to prepare fluorescent enzyme substrates, fatty acids, phospholipids, lipopolysaccharides, receptor ligands and polystyrene microspheres.
Also used for solid-state dye laser devices and organic solar cells.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Iyappan Ramachandiran et al.
Scientific reports, 8(1), 1154-1154 (2018-01-20)
Protein phosphorylation is an important post-translational modification that can regulate the protein function. The current knowledge on the phosphorylation status of plant oil body (OB) proteins is inadequate. This present study identifies the distinct physiological substrates of Arabidopsis serine/threonine/tyrosine protein
Sameh Rabhi et al.
PloS one, 11(2), e0148640-e0148640 (2016-02-13)
Leishmania, the causative agent of vector-borne diseases, known as leishmaniases, is an obligate intracellular parasite within mammalian hosts. The outcome of infection depends largely on the activation status of macrophages, the first line of mammalian defense and the major target
BODIPY dyes and their derivatives: syntheses and spectroscopic properties.
Loudet A and Burgess K
Chemical Reviews, 107(11), 4891-4932 (2007)
Infused polymers for cell sheet release.
Juthani N, et al.
Scientific Reports, 6(11), 26109-26109 (2016)
Inhibition of the acetyl lysine-binding pocket of bromodomain and extraterminal domain proteins interferes with adipogenesis.
Goupille O, et al.
Biochemical and Biophysical Research Communications, 472(4), 624-630 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service