Skip to Content
Merck
All Photos(1)

Documents

735132

Sigma-Aldrich

Allylamine hydrochloride

98%

Synonym(s):

3-Aminopropene hydrochloride, Mono-allylamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H7N · HCl
CAS Number:
Molecular Weight:
93.56
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

98%

form

solid

mp

178-183 °C (dec.)

SMILES string

Cl.NCC=C

InChI

1S/C3H7N.ClH/c1-2-3-4;/h2H,1,3-4H2;1H

InChI key

MLGWTHRHHANFCC-UHFFFAOYSA-N

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aqiang Wang et al.
Small (Weinheim an der Bergstrasse, Germany), 16(9), e1903925-e1903925 (2019-10-11)
Highly sensitive responsiveness is vital for stimuli-responsive membranes. However, it is a great challenge to fabricate stimuli-responsive membranes with ultrahigh gating ratio (the ratio of the salt solution permeating flux to the pure water permeating flux) and high response speed
Jianlong Wang et al.
Langmuir : the ACS journal of surfaces and colloids, 35(10), 3615-3623 (2019-02-13)
Interfacial gas enrichment (IGE) of dissolved gases in water is shown to govern the strong attraction between solid hydrophobic surfaces of an atomic force microscopy (AFM) colloidal probe and solid substrate. However, the role of IGE in controlling the attraction
Lu Han et al.
Biomaterials, 44, 111-121 (2015-01-27)
Multifunctional nanocomplexes (NCs) consisting of urocanic acid-modified galactosylated trimethyl chitosan (UA-GT) conjugates as polymeric vectors, poly(allylamine hydrochloride)-citraconic anhydride (PAH-Cit) as charge-reversible crosslinkers, and vascular endothelial growth factor (VEGF) siRNA as therapeutic genes, were rationally designed to simultaneously overcome the extracellular
Wei Li et al.
Biomaterials, 65, 93-102 (2015-07-06)
Selective isolation and purification of circulating tumor cells (CTCs) from whole blood is an important capability for both clinical medicine and biological research. Current techniques to perform this task place the isolated cells under excessive stresses that reduce cell viability
Seyeong Lee et al.
Nanoscale, 7(35), 14627-14635 (2015-07-23)
Recent advances in nanostructure-based biotechnology have resulted in a growing demand for vertical nanostructure substrates with elaborate control over the nanoscale geometry and a high-throughput preparation. In this work, we report the fabrication of non-periodic vertical silicon nanocolumn substrates via

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service