Skip to Content
Merck
All Photos(2)

Documents

P50900

Sigma-Aldrich

Propargylamine

98%

Synonym(s):

2-Propynylamine, 3-Amino-1-propyne

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HC≡CCH2NH2
CAS Number:
Molecular Weight:
55.08
Beilstein:
773681
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

contains

≤2.5% water

refractive index

n20/D 1.449 (lit.)

bp

83 °C (lit.)

density

0.86 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

NCC#C

InChI

1S/C3H5N/c1-2-3-4/h1H,3-4H2

InChI key

JKANAVGODYYCQF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Synthesis of a chiral, fluorescent macrocycle by 1,3-dipolar cycloaddition of propargyl amides of carbohydrate-linked amino acids and 9,10-bis(azidomethyl)anthracene.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

42.8 °F - closed cup

Flash Point(C)

6 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, 3141-3141 (2006)
Claudio Gnaccarini et al.
Organic & biomolecular chemistry, 10(27), 5258-5265 (2012-06-02)
Transglutaminases (TGases) catalyse the transamidation of glutamine residues with primary amines. Herein we report the first FRET-based activity assay for the direct detection of the ligation (transamidation) reaction mediated by tissue TGase (TG2). This novel assay was then used in
Anna Fodor et al.
Organic & biomolecular chemistry, 8(20), 4575-4581 (2010-08-27)
A new, heterogeneous, 4 A molecular sieve-supported copper(ii) catalyst was developed and was used successfully in the A(3) coupling of alkynes, aldehydes and amines under simple reaction conditions.
Adrian Sulistio et al.
Chemical communications (Cambridge, England), 47(4), 1151-1153 (2010-12-15)
Highly functionalized water soluble core cross-linked star (CCS) polymers having degradable cores and hierarchical functionalities spanning from the peripheral groups along the arms to the core have been synthesized entirely from amino acid building blocks. The core-isolated moieties were shown
Irene Bolea et al.
Journal of neural transmission (Vienna, Austria : 1996), 120(6), 893-902 (2012-12-15)
Alzheimer's disease (AD) is a complex neurodegenerative disorder with a multifaceted pathogenesis. There are at present three Food and Drug Administration-approved drugs based on the "one drug, one target" paradigm (donepezil, galantamine and rivastigmine) that improve symptoms by inhibiting acetylcholinesterase.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service