Skip to Content
Merck
All Photos(1)

Key Documents

CDS000416

Sigma-Aldrich

2-Propen-1-amine hydrochloride

AldrichCPR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H8ClN
CAS Number:
Molecular Weight:
93.56
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

form

solid

SMILES string

Cl.NCC=C

InChI

1S/C3H7N.ClH/c1-2-3-4;/h2H,1,3-4H2;1H

InChI key

MLGWTHRHHANFCC-UHFFFAOYSA-N

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lu Han et al.
Biomaterials, 44, 111-121 (2015-01-27)
Multifunctional nanocomplexes (NCs) consisting of urocanic acid-modified galactosylated trimethyl chitosan (UA-GT) conjugates as polymeric vectors, poly(allylamine hydrochloride)-citraconic anhydride (PAH-Cit) as charge-reversible crosslinkers, and vascular endothelial growth factor (VEGF) siRNA as therapeutic genes, were rationally designed to simultaneously overcome the extracellular
Wei Li et al.
Biomaterials, 65, 93-102 (2015-07-06)
Selective isolation and purification of circulating tumor cells (CTCs) from whole blood is an important capability for both clinical medicine and biological research. Current techniques to perform this task place the isolated cells under excessive stresses that reduce cell viability
Seyeong Lee et al.
Nanoscale, 7(35), 14627-14635 (2015-07-23)
Recent advances in nanostructure-based biotechnology have resulted in a growing demand for vertical nanostructure substrates with elaborate control over the nanoscale geometry and a high-throughput preparation. In this work, we report the fabrication of non-periodic vertical silicon nanocolumn substrates via
Lei Wu et al.
Small (Weinheim an der Bergstrasse, Germany), 11(23), 2798-2806 (2015-02-18)
A surface enhanced Raman scattering (SERS)-assisted 3D barcode chip has been developed for high-throughput biosensing. The 3D barcode is realized through joint 2D spatial encoding with the Raman spectroscopic encoding, which stores the SERS fingerprint information in the format of
Chunli Zheng et al.
International journal of pharmaceutics, 475(1-2), 17-24 (2014-08-28)
In this study, magneto-responsive polyelectrolyte multilayer microcapsules were successfully prepared by the formation of shell with biocompatible iron oxide nanoparticles (Fe₃O₄ NPs) and polyallylamine hydrochloride (PAH) by layer-by-layer (LbL) self-assembly technique. The self-assembled microcapsules were characterized by SEM, TEM and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service