Skip to Content
Merck
All Photos(3)

Documents

731625

Sigma-Aldrich

trans-1,2-Bis(tributylstannyl)ethene

97%

Synonym(s):

(E)-1,2-Bis(tributylstannyl)ethane, (E)-1,2-Bis(tributylstannyl)ethylene, (E)-1,2-Bis(tributyltin)ethylene, (E)-1,2-Ethenediylbis[tributylstannane], (E)-Bis(tributylstannyl)ethene, 1,2-Ethenediylbis[tributylstannane], trans-1,2-Bis(tributylstannyl)ethylene, trans-Bis(tributylstannyl)ethene, trans-Bis(tributylstannyl)ethylene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H56Sn2
CAS Number:
Molecular Weight:
606.14
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.502

density

1.132 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CCCC[Sn](CCCC)(CCCC)\C=C\[Sn](CCCC)(CCCC)CCCC

InChI

1S/6C4H9.C2H2.2Sn/c6*1-3-4-2;1-2;;/h6*1,3-4H2,2H3;1-2H;;

InChI key

VNKOWRBFAJTPLS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

trans-1,2-Bis(tributylstannyl)ethene can be used to prepare:
  • Polyarylenevinylene polymers having triphenylamine core moieties.
  • Two (E)-2-(2-(thiophen-2-yl)vinyl)thiophen based copolymers through Pd-catalyzed Stille coupling reaction.
  • Vinylstannanes by reacting with aryl halides, which in turn can be used in the synthesis of substituted pyridines.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tuning optical and electronic properties of poly (4, 4′-triphenylamine vinylene) s by post-modification reactions
Grigoras M, et al.
Dyes and Pigments, 113, 227-238 (2015)
Synthesis, characterization, and field-effect properties of (E)-2-(2-(thiophen-2-yl) vinyl) thiophen-based donor-acceptor copolymers
Wang L, et al.
Polymer, 68, 302-307 (2015)
A novel strategy for the synthesis of 2-arylpyridines using one-pot 6π-azaelectrocyclization
Kobayashi T, et al.
Tetrahedron Letters, 49(28), 4349-4351 (2008)
Brigitte Holzer et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 26(13), 2869-2882 (2019-11-16)
A reliable synthetic protocol toward a series of fused chalcogenopheno[1]benzochalcogenophene (CBC) building blocks was developed based on a Fiesselmann reaction. The obtained CBC units were applied in McMurry and Stille coupling reactions toward symmetric regioisomeric ene-linked dimers. These π-conjugated compounds

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service