Skip to Content
Merck
All Photos(1)

Key Documents

613223

Supelco

Nicotine solution

NMR reference standard, 20 mg/mL in acetone-d6 (99.9 atom % D), TMS 0.05 %, CFCl3 0.4 %, NMR tube size 5 mm × 8 in.

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14N2
Molecular Weight:
162.23
UNSPSC Code:
12142201
NACRES:
NA.24
Pricing and availability is not currently available.

grade

NMR reference standard
analytical standard

composition

CFCl3, 0.4%
TMS, 0.05%

concentration

20 mg/mL in acetone-d6 (99.9 atom % D)

technique(s)

NMR: suitable

NMR tube size

5 mm × 8 in.

suitability

suitable for NMR (reference standard)

format

single component solution

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

NMR reference standard (20 mg/mL in acetone-d6 (99.9 atom % D), TMS 0.05%, CFCl3 0.4%) eliminates the sample preparation uncertainty when evaluating instrument performance and assures proper instrument performance in quantitative applications. These NMR reference standards allow the users to monitor key operational parameters such as pw90, sensitivity, resolution, and line shape.

Quantity

5 mm O.D. tube contains 0.700 mL.
Each pack contains 10 tubes.

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Ozone 1 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-2.2 °F

Flash Point(C)

-19 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service