Skip to Content
Merck
All Photos(1)

Key Documents

36733

Supelco

(−)-Nicotine

PESTANAL®, analytical standard

Synonym(s):

(−)-1-Methyl-2-(3-pyridyl)pyrrolidine, (S)-3-(1-Methyl-2-pyrrolidinyl)pyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14N2
CAS Number:
Molecular Weight:
162.23
Beilstein:
82109
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.5265 (lit.)

density

1.010 g/mL at 20 °C (lit.)

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

CN1CCC[C@H]1c2cccnc2

InChI

1S/C10H14N2/c1-12-7-3-5-10(12)9-4-2-6-11-8-9/h2,4,6,8,10H,3,5,7H2,1H3/t10-/m0/s1

InChI key

SNICXCGAKADSCV-JTQLQIEISA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Prototype nicotinic acetylcholine receptor agonist; naturally occurring isomer.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Oral - Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

213.8 °F - closed cup

Flash Point(C)

101 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Ramkrishna Mehendale et al.
American journal of obstetrics and gynecology, 197(4), 363-363 (2007-10-02)
Excess soluble vascular endothelial growth factor receptor, fms-like tyrosine kinase-1 (sFlt-1), and reduced placental growth factor (PlGF) mediate the genesis of preeclampsia. Cigarette smoking reduces the risk of preeclampsia. We hypothesized that placental secretion of sFlt-1 and PlGF was affected
Charles C Horn et al.
American journal of physiology. Gastrointestinal and liver physiology, 298(1), G25-G30 (2009-11-07)
Nausea and emesis are common side effects of gastrointestinal disease. Reports indicate that ghrelin and endocannabinoids, agents that stimulate appetite, also reduce emesis evoked by chemotherapy treatment, which suggests that stimulation of feeding inhibits the emetic system. In the following
Pooja Naik et al.
BMC neuroscience, 15, 51-51 (2014-04-24)
Both active and passive tobacco smoke (TS) potentially impair the vascular endothelial function in a causative and dose-dependent manner, largely related to the content of reactive oxygen species (ROS), nicotine, and pro-inflammatory activity. Together these factors can compromise the restrictive
J A Hutcherson et al.
Molecular oral microbiology, 31(4), 354-364 (2015-09-12)
Porphyromonas gingivalis is a Gram-negative anaerobe and keystone periodontal pathogen. A mariner transposon insertion mutant library has recently been used to define 463 genes as putatively essential for the in vitro growth of P. gingivalis ATCC 33277 in planktonic culture (Library 1). We
Charles C Horn et al.
American journal of physiology. Regulatory, integrative and comparative physiology, 306(5), R341-R351 (2014-01-17)
Signals from the vestibular system, area postrema, and forebrain elicit nausea and vomiting, but gastrointestinal (GI) vagal afferent input arguably plays the most prominent role in defense against food poisoning. It is difficult to determine the contribution of GI vagal

Protocols

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Questions

1–2 of 2 Questions  
  1. What's the concentration? 1 mg/mL??

    1 answer
    1. This product is a neat or pure liquid with a density of 1.01 g/mL. The minimum purity is ~98% by GC analysis, as listed on the lot specific Certificate of Analysis. Please navigate to the ‘DOCUMENTATION’ section of the Product Detail Page to access a sample of lot specific certificate:
      https://www.sigmaaldrich.com/product/sial/36733#product-documentation

      Helpful?

  2. Can stock dilutions in DI water be stored at -20C long term? If so, how long can the solutions be stored?

    1 answer
    1. Unfortunately, stability information for Product 36733 is not determined for the reconstituted material. Due to this, freezing the stock solution and keeping track of the stability of the solution is at the discretion and liberty of the end-user.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service