Yes, this product is a neat or pure form of Nicotine. Based on a density of 1.02 g/ml and a molecular weight of 162.2 g/mole, this neat liquid has a calculated molar concentration of 6.3 M.
8.20877
(-)-Nicotine
for synthesis
Synonym(s):
(-)-Nicotine, 3-(1-Methyl-2-pyrrolidinyl)pyridine, (S)-(-)-Nicotine
About This Item
Recommended Products
vapor pressure
0.06 hPa ( 20 °C)
Quality Level
Assay
≥99.0% (GC)
form
liquid
autoignition temp.
240 °C
potency
50 mg/kg LD50, oral (Rat)
50 mg/kg LD50, skin (Rabbit)
expl. lim.
0.7-4 % (v/v)
pH
10.2 (20 °C, 8.1 g/L in H2O)
bp
248.8 °C/1013 hPa
mp
-79 °C
transition temp
flash point 111 °C
density
1.01 g/cm3 at 20 °C
storage temp.
2-30°C
SMILES string
N1([C@@H](CCC1)c2cnccc2)C
InChI
1S/C10H14N2/c1-12-7-3-5-10(12)9-4-2-6-11-8-9/h2,4,6,8,10H,3,5,7H2,1H3/t10-/m0/s1
InChI key
SNICXCGAKADSCV-JTQLQIEISA-N
Analysis Note
Density (d 20 °C/ 4 °C): 1.009 - 1.012
Enantiomeric purity: ≥ 99.0 % (a/a)
Identity (IR): passes test
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 1 Oral - Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2
Storage Class Code
6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK
WGK 3
Flash Point(F)
213.8 °F - closed cup
Flash Point(C)
101 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
This is pure nicotine solution, right? This is not nicotine that has been dissolved in a solvent.
1 answer-
Helpful?
-
-
What is the concentration in the 25ml bottle preferably in mg/ml
1 answer-
This product is a neat liquid with a minimum purity of 99%. The density is 1.01 g/mL and the molecular weight is 162.23. The calculated concentration of this product is approximately 6.2 M. This value will change slightly when considering the lot specific purity. This product is not tested for moisture content.
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service