Skip to Content
Merck
All Photos(1)

Key Documents

516155

Sigma-Aldrich

2-Aminoethyl methacrylate hydrochloride

contains ~500 ppm phenothiazine as stabilizer, 90%

Synonym(s):

2-Aminoethyl 2-methylacrylate hydrochloride, 2-Propenoic acid, 2-methyl-, 2-aminoethyl ester, hydrochloride (1:1) (ACI)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=C(CH3)CO2CH2CH2NH2 · HCl
CAS Number:
Molecular Weight:
165.62
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

90%

contains

~500 ppm phenothiazine as stabilizer

mp

102-110 °C (lit.)

storage temp.

2-8°C

SMILES string

Cl.CC(=C)C(=O)OCCN

InChI

1S/C6H11NO2.ClH/c1-5(2)6(8)9-4-3-7;/h1,3-4,7H2,2H3;1H

InChI key

XSHISXQEKIKSGC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Aminoethyl methacrylate hydrochloride (AMA) is an amine based methacrylic monomer used in the production of polymers and copolymers that are utilized in a wide range of applications, including coatings, adhesives, and medical devices. It is a highly reactive monomer due to the presence of the methacrylate group, which enables it to undergo both free radical polymerization and other polymerization reactions. Due to its biocompatibility and low toxicity, it is widely used in biomedical research and as a crosslinking agent for hydrogels and other biomaterials. It is used in the synthesis of poly (2-aminoethylmethacrylate), a biocompatible cationic polymer widely used in the biomedical field.

Application

2-Aminoethylmethacrylate hydrochloride(AMA) can be used as a precursor to synthesize:
  • Homo and copolymer brushes of AMA, which can be used to tune the electrochemical properties of silicon wafers.
  • Poly(3-hydroxybutyrate) based polyurethane/urea composite scaffolds for tissue engineering applications.
  • Antimicrobial bacterial cellulose/poly (AMA) nanocomposites. Polymer incorporation imparts mechanical properties and antimicrobial activity to bacterial cellulose membranes.
It can also be used for the efficient functionalization of hyaluronic acid via triazine-mediated amidation.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Poly (ethylene glycol)-block-poly (2-aminoethyl methacrylate hydrochloride)-Based Polyplexes as Serum-Tolerant Nanosystems for Enhanced Gene Delivery
Santo D, et al.
Molecular Pharmaceutics, 16, 2129-2141 (2019)
Mario Lopez-Moya et al.
Biomacromolecules, 19(4), 1234-1244 (2018-03-15)
Porcine glutaraldehyde-fixed pericardium is widely used to replace human heart valves. Despite the stabilizing effects of glutaraldehyde fixation, the lack of endothelialization and the occurrence of immune reactions contribute to calcification and structural valve deterioration, which is particularly significant in
Gaëlle Levourch et al.
Polymers, 12(6) (2020-07-01)
In biomedical diagnosis and bionanotechnologies, the extraction and purification of proteins and protein derivatives are of great interest. In fact, to purify recombinant proteins for instance, new methodologies and well appropriate material supports need to be established and also to
Aminomalononitrile-assisted multifunctional antibacterial coatings
Liao T-Y, et al.
ACS biomaterials science & engineering, 6, 3349-3360 (2020)
Ana Margarida Piloto et al.
Scientific reports, 8(1), 4944-4944 (2018-03-23)
A highly sensitive fluorescence detection probe was developed by tailoring plastic antibodies on the external surface of aqueous soluble quantum dots (QDs). The target was Myoglobin (Myo), a cardiac biomarker that quenched the intrinsic fluorescent emission of cadmium telluride (CdTe)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service