Skip to Content
Merck
All Photos(2)

Key Documents

408972

Sigma-Aldrich

2-(Diethylamino)ethyl acrylate

95%, contains 25 ppm monomethyl ether hydroquinone as inhibitor

Synonym(s):

(N ,N -Diethylamino)ethyl acrylate, [2-(Acryloyloxy)ethyl]diethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCOOCH2CH2N(C2H5)2
CAS Number:
Molecular Weight:
171.24
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

0.77 psi ( 20 °C)

Assay

95%

contains

25 ppm monomethyl ether hydroquinone as inhibitor

refractive index

n20/D 1.443 (lit.)

bp

199 °C (lit.)

density

0.922 g/mL at 25 °C (lit.)

SMILES string

CCN(CC)CCOC(=O)C=C

InChI

1S/C9H17NO2/c1-4-9(11)12-8-7-10(5-2)6-3/h4H,1,5-8H2,2-3H3

InChI key

QHVBLSNVXDSMEB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

165.0 °F - closed cup

Flash Point(C)

73.9 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yujing You et al.
Journal of biomedical materials research. Part B, Applied biomaterials, 107(6), 1786-1791 (2018-11-16)
Cell growth and release from the hydrogel scaffolds are critical steps in cell culture and cell therapies. Herein a double network hydrogel with one thermosensitive P(AEtMA-Cl-DEAEA) network and the other polyacrylamide network is reported. The diad hydrogel is elastic and
Mayra Eliana Valencia Zapata et al.
Polymers, 12(7) (2020-07-28)
Despite the potential of acrylic bone cement (ABC) loaded with chitosan (CS) for orthopedic applications, there are only a few in vitro studies of this composite with CS loading ≤ 15 wt.% evaluated in bioactivity tests in simulated body fluid
Nasha Qiu et al.
Biomacromolecules, 19(6), 2308-2319 (2018-05-09)
Gene therapy has demonstrated effectiveness in many genetic diseases, as evidenced by recent clinical applications. Viral vectors have been extensively tested in clinical gene-therapy trials, but nonviral vectors such as cationic polymers or lipids are much less used due to
Robert Löwe et al.
Molecules (Basel, Switzerland), 24(2) (2019-01-20)
Eight new polymerizable ammonium-TFSI ionic liquids were synthesized and characterized with respect to an application in energy storage devices. The ionic liquids feature methacrylate or acrylate termination as polymerizable groups. The preparation was optimized to obtain the precursors and ionic
Lucia Cenci et al.
Journal of nanobiotechnology, 13, 51-51 (2015-08-28)
Molecularly imprinted polymer (MIP) technique is a powerful mean to produce tailor made synthetic recognition sites. Here precipitation polymerization was exploited to produce a library of MIP nanoparticles (NPs) targeting the N terminus of the hormone Hepcidin-25, whose serum levels

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service