Skip to Content
Merck
All Photos(3)

Key Documents

477060

Sigma-Aldrich

2-Isocyanatoethyl methacrylate

contains ≤0.1% BHT as inhibitor, 98%

Synonym(s):

2-(Methacryloyloxy)ethyl isocyanate, IEM

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=C(CH3)CO2CH2CH2NCO
CAS Number:
Molecular Weight:
155.15
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

contains

≤0.1% BHT as inhibitor

refractive index

n20/D 1.45 (lit.)

bp

211 °C (lit.)

mp

−45 °C (lit.)

density

1.098 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(=C)C(=O)OCCN=C=O

InChI

1S/C7H9NO3/c1-6(2)7(10)11-4-3-8-5-9/h1,3-4H2,2H3

InChI key

RBQRWNWVPQDTJJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Isocyanatoethyl methacrylate, belongs to the class of monomers known as isocyanate monomers. The presence of the isocyanate functional group in 2-Isocyanatoethyl methacrylate makes it highly reactive. This reactivity allows it to undergo polymerization and form cross-linked networks for the development of polymers with improved mechanical strength, chemical resistance, and adhesion properties. It is also used as a crosslinker in the manufacture of many resins, in electrodeposition, and adhesives, and as a co-monomer in crosslinked polymer polyolstyrene-acrylic resin, which is used in construction products, such as ceramic tile adhesives, fillers, putties, and elastomeric roof coatings.

Application

2-Isocyanatoethyl methacrylate can be used as:
  • A reactive monomer in the synthesis of poly(methyl urethane) acrylate oligomer for UV-curable coatings. By incorporating urethane and acrylate groups into the polymer, it enables the polymer to be utilized in UV-curable coatings, inks, and adhesives.
  • An additive in propylene carbonate-based electrolyte to enhance its properties, such as viscosity, conductivity, and stability. This improves the overall performance of the electrolyte and the battery.
  • As a modifier in the copolymerization process to prepare a bioprosthetic heart valve, which is used in the application of transcatheter aortic valve replacement.
  • A latent cross-linker for adhesive resins and coatings.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

210.2 °F - closed cup

Flash Point(C)

99 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Cock et al.
Neurogastroenterology and motility : the official journal of the European Gastrointestinal Motility Society, 28(12), 1890-1901 (2016-06-28)
Dysphagia becomes more common in old age. We performed high-resolution impedance manometry (HRIM) in asymptomatic healthy adults (including an older cohort >80 years) to assess HRIM findings in relation to bolus clearance. Esophageal HRIM was performed in a sitting posture in
Sheng H Wang et al.
NeuroImage, 173, 610-622 (2018-01-30)
Inter-areal functional connectivity (FC), neuronal synchronization in particular, is thought to constitute a key systems-level mechanism for coordination of neuronal processing and communication between brain regions. Evidence to support this hypothesis has been gained largely using invasive electrophysiological approaches. In
Clare Williams et al.
Nutrition & dietetics: the journal of the Dietitians Association of Australia, 76(2), 141-149 (2019-03-09)
This project aimed to implement consensus recommendations and innovations that improve dietetic services to promote timely referral to optimise nutritional management for adult inpatients with inborn errors of metabolism (IEM). The i-PARIHS framework was used to identify service gaps, implement
Cranley, P.E.
Polym. Sci. Technol., 29, 765-765 (1984)
Bingxian Yang et al.
Journal of proteomics, 150, 323-340 (2016-10-22)
High level of UV-B irradiation followed by dark treatment (HUV-B+D) causes accumulation of secondary metabolites in Clematis terniflora DC. To investigate the response mechanism under HUV-B+D, transcriptomic and proteomic analyses were performed in leaves of C. terniflora. The number of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service