Skip to Content
Merck
All Photos(1)

Key Documents

447153

Sigma-Aldrich

trans-3-Hexene

≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5CH=CHC2H5
CAS Number:
Molecular Weight:
84.16
Beilstein:
1718859
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥99%

refractive index

n20/D 1.394 (lit.)

bp

67 °C (lit.)

density

0.677 g/mL at 25 °C (lit.)

SMILES string

CC\C=C\CC

InChI

1S/C6H12/c1-3-5-6-4-2/h5-6H,3-4H2,1-2H3/b6-5+

InChI key

ZQDPJFUHLCOCRG-AATRIKPKSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-13.0 °F - closed cup

Flash Point(C)

-25 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and ene reactions of di-(-)-menthyl diazenedicarboxylate.
Brimble MA, et al.
Tetrahedron Asymmetry, 7(7), 2007-2016 (1996)
Study on the separation of 1-hexene and trans-3-hexene using ionic liquids.
Jiqin Z, et al.
Fluid Phase Equilibria, 247(1), 102-106 (2006)
Electronic effects of bis (2-aryl-4, 5, 6, 7-tetrahydroindenyl) titanocene dichlorides on the catalytic epoxidation of trans-3-hexene.
Halterman RL and Ramsey TM.
Journal of Organometallic Chemistry, 465(1), 175-179 (1994)
Oligomers from the ozonolyses of cis-and trans-3-hexene and cis-and trans-2, 5-dimethyl-3-hexene
Murray RW and Su JS.
The Journal of Organic Chemistry, 48(6), 817-822 (1983)
Alla Zelenyuk et al.
Faraday discussions, 200, 143-164 (2017-06-06)
When secondary organic aerosol (SOA) particles are formed by ozonolysis in the presence of gas-phase polycyclic aromatic hydrocarbons (PAHs), their formation and properties are significantly different from SOA particles formed without PAHs. For all SOA precursors and all PAHs, discussed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service