Skip to Content
Merck
All Photos(3)

Documents

413526

Sigma-Aldrich

2,2′-Diallylbisphenol A

technical grade, 85%

Synonym(s):

2,2′-Bis(3-allyl-4-hydroxyphenyl)isopropylidene, 2,2-Bis(3-allyl-4-hydroxyphenyl)propane, 4,4′-(1-Methylethylidene)bis[2-(2-propenyl)phenol], 4,4′-Isopropylidenebis(2-allylphenol)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C[C6H3(CH2CH=CH2)OH]2
CAS Number:
Molecular Weight:
308.41
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

grade

technical grade

Quality Level

Assay

85%

form

viscous liquid

impurities

<2% ethyl acetate

refractive index

n20/D 1.587 (lit.)

density

1.08 g/mL at 25 °C (lit.)

SMILES string

CC(C)(c1ccc(O)c(CC=C)c1)c2ccc(O)c(CC=C)c2

InChI

1S/C21H24O2/c1-5-7-15-13-17(9-11-19(15)22)21(3,4)18-10-12-20(23)16(14-18)8-6-2/h5-6,9-14,22-23H,1-2,7-8H2,3-4H3

InChI key

WOCGGVRGNIEDSZ-UHFFFAOYSA-N

General description

2,2′-Diallylbisphenol A (DABA) is a diallyl based epoxy modification agent that blends with the resin to improve the mechanical property of the epoxy material. It is majorly used in novalac based resins, which can be further cured with bismaleimides (BMI) for aerospace, electronics and wireless communication based applications.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cure reaction for modified diallylbisphenol A/diaminodiphenylsulfone/bismaleimide
Boey FYC, et al.
Journal of Applied Polymer Science, 85(2), 227-235 (2002)
Diallyl bisphenol A?novolac epoxy system cocured with bisphenol-A-bismaleimide?cure and thermal properties
Ambika DK, et al.
Journal of Applied Polymer Science, 106(2), 1192-1200 (2007)
Catalytic effect of 2, 2′-diallyl bisphenol A on thermal curing of cyanate esters
Koh HCY, et al.
Journal of Applied Polymer Science, 101(3), 1775-1786 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service