Skip to Content
Merck
All Photos(2)

Key Documents

405981

Sigma-Aldrich

2,2′-Methylenebis[(4R,5S)-4,5-diphenyl-2-oxazoline]

99%

Synonym(s):

(4R,5S,4′R,5′S)-2,2′-Methylenebis(4,5-diphenyl-2-oxazoline)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H26N2O2
CAS Number:
Molecular Weight:
458.55
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

mp

205-208 °C (lit.)

SMILES string

C(C1=N[C@@H]([C@@H](O1)c2ccccc2)c3ccccc3)C4=N[C@@H]([C@@H](O4)c5ccccc5)c6ccccc6

InChI

1S/C31H26N2O2/c1-5-13-22(14-6-1)28-30(24-17-9-3-10-18-24)34-26(32-28)21-27-33-29(23-15-7-2-8-16-23)31(35-27)25-19-11-4-12-20-25/h1-20,28-31H,21H2/t28-,29-,30+,31+/m1/s1

InChI key

BVEHHQYXICTXGR-VKONIRKNSA-N

Application

C2 symmetric ligand for enantioselective catalysis. Easily forms bidentate coordination complexes due to the strong affinity of the oxazoline nitrogen for various metals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bolm, C.
Angewandte Chemie (International Edition in English), 30, 542-542 (1991)
Gant, T.G. Meyers, A.I.
Tetrahedron, 50, 2297-2297 (1994)
Pfaltz, A.
Accounts of Chemical Research, 26, 339-339 (1993)

Articles

BOX ligand complex enhances catalytic enantioselective aziridination of styrene, with phenyl substituents proving superior over t-Bu groups.

BOX ligand complex enhances catalytic enantioselective aziridination of styrene, with phenyl substituents proving superior over t-Bu groups.

BOX ligand complex enhances catalytic enantioselective aziridination of styrene, with phenyl substituents proving superior over t-Bu groups.

BOX ligand complex enhances catalytic enantioselective aziridination of styrene, with phenyl substituents proving superior over t-Bu groups.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service