Skip to Content
Merck
All Photos(3)

Key Documents

441791

Sigma-Aldrich

Tris(2,4-di-tert-butylphenyl) phosphite

Synonym(s):

2,4-Bis(1,1-dimethylethyl)phenol phosphite (3:1), Tri(2,4-di-tert -butylphenyl) phosphite, Tri(2,4-di-t -butylphenyl) phosphite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[[(CH3)3C]2C6H3O]3P
CAS Number:
Molecular Weight:
646.92
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

mp

181-184 °C (lit.)

SMILES string

CC(C)(C)c1ccc(OP(Oc2ccc(cc2C(C)(C)C)C(C)(C)C)Oc3ccc(cc3C(C)(C)C)C(C)(C)C)c(c1)C(C)(C)C

InChI

1S/C42H63O3P/c1-37(2,3)28-19-22-34(31(25-28)40(10,11)12)43-46(44-35-23-20-29(38(4,5)6)26-32(35)41(13,14)15)45-36-24-21-30(39(7,8)9)27-33(36)42(16,17)18/h19-27H,1-18H3

InChI key

JKIJEFPNVSHHEI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tris(2,4-di-tert-butylphenyl) phosphite is a triaryl based phosphite that can be used in catalysis and metallation. Its characteristic to undergo metallation reaction and provide a cost effective synthetic processes allows it to be useful in biaryl coupling reactions.

Application

It may be used as a chain extender and a processing stabilizer that facilitate the production of stable polymeric materials, which include polylactic acid (PLA), polyolefins, etc.

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Orthopalladated triarylphosphite complexes as highly efficient catalysts in the Heck reaction.
Albisson DA
Tetrahedron Letters, 39(52), 9793-9796 (1998)
Orthopalladated triaryl phosphite complexes as highly active catalysts in biaryl coupling reactions.
Albisson DA, et al.
Chemical Communications (Cambridge, England), 19, 2095-2096 (1998)
Study of the high temperature reactions of a hindered aryl phosphite (Hostanox PAR 24) used as a processing stabiliser in polyolefins.
Kriston I, et al.
Polymer Degradation and Stability, 95(9), 1883-1893 (2010)
Structure effect of phosphite on the chain extension in PLA.
Meng X, et al.
Polymer Degradation and Stability, 120(52), 283-289 (2015)
Taoufiq Mouhib et al.
Journal of the American Society for Mass Spectrometry, 21(12), 2005-2010 (2010-09-25)
The enhancement of the static secondary ion mass spectrometry (SIMS) signals resulting from the injection, closely to the sample surface, of H(2)O vapor at relatively high-pressure, was investigated for a set of organic materials. While the ion signals are generally

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service