Skip to Content
Merck
All Photos(1)

Documents

773212

Sigma-Aldrich

N,N-Diethylacrylamide

contains <200 ppm MEHQ as inhibitor, 99%

Synonym(s):

N,N-Diethyl-2-propenamide, Acrylic acid diethylamide, DEAA, DEAAm, DEAM

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H13NO
CAS Number:
Molecular Weight:
127.18
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

form

liquid

contains

<200 ppm MEHQ as inhibitor

refractive index

n20/D 1.468

density

0.924 at 25 °C

storage temp.

2-8°C

SMILES string

CCN(CC)C(=O)C=C

InChI

1S/C7H13NO/c1-4-7(9)8(5-2)6-3/h4H,1,5-6H2,2-3H3

InChI key

OVHHHVAVHBHXAK-UHFFFAOYSA-N

Application

Acrylamide monomer used to make polymers for drug delivery; thermal responsive polymers; and solutions with specifically tuned LCSTs.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

192.0 °F

Flash Point(C)

88.89 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nuran Işıklan et al.
International journal of biological macromolecules, 113, 669-680 (2018-03-03)
The functionalization of polysaccharides with synthetic polymers has attracted great attention owing to its application in many industrial fields. The aim of this work was to study the impact of pectin functionalization with N,N-diethylacrylamide (DEAAm). Pectin was modified via microwave-induced
Z Ding et al.
Nature, 411(6833), 59-62 (2001-05-03)
Many medical and biotechnological processes rely on controlling and manipulating the molecular-recognition capabilities of proteins. This can be achieved using small molecules capable of competing for protein binding or by changing environmental parameters that affect protein structure and hence binding.
Wei Wei et al.
Colloids and surfaces. B, Biointerfaces, 136, 1182-1192 (2015-11-23)
Salecan is a water-soluble microbial polysaccharide produced by Agrobacterium sp. ZX09, a salt-tolerant strain isolated from a soil sample in our laboratory. Previous work inspired us salecan is a good candidate to fabricate hydrogels. Poly(N,N-diethylacrylamide) is one type of thermo
Xuemei Wu et al.
Sensors (Basel, Switzerland), 18(10) (2018-10-10)
In this work, binary hydrogel films based on carboxylated multi-walled carbon nanotubes/poly(N,N-diethylacrylamide) (c-MWCNTs/PDEA) were successfully polymerized and assembled on a glassy carbon (GC) electrode surface. The electroactive drug probes matrine and sophoridine in solution showed reversible thermal-, salt-, methanol- and
Seog-Jin Jeon et al.
Soft matter, 16(3), 688-694 (2019-12-10)
Trilayer polymer films consisting of a thermoresponsive hydrogel, poly(diethyl acrylamide) (PDEAM), sandwiched by rigid layers of a glassy polymer, poly(para-methylstyrene) (PpMS), patterned into parallel striped features are prepared and used to drive temperature-responsive reversible anisotropic expansion. Significant swelling occurs along

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service