Skip to Content
Merck
All Photos(1)

Key Documents

40225

Sigma-Aldrich

Dimethylditetradecylammonium bromide

≥97.0% (NT)

Synonym(s):

Ditetradecyldimethylammonium bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)13]2N(Br)(CH3)2
CAS Number:
Molecular Weight:
518.74
Beilstein:
4050300
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0% (NT)

mp

167-169 °C (lit.)

SMILES string

[Br-].CCCCCCCCCCCCCC[N+](C)(C)CCCCCCCCCCCCCC

InChI

1S/C30H64N.BrH/c1-5-7-9-11-13-15-17-19-21-23-25-27-29-31(3,4)30-28-26-24-22-20-18-16-14-12-10-8-6-2;/h5-30H2,1-4H3;1H/q+1;/p-1

InChI key

IRMGVPILCPGYNQ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Dimethylditetradecylammonium bromide can be used:
  • To coat the capillary column used in the capillary electrophoresis of protein-dye complexes.
  • To prepare Fe-porphyrin based pH-sensitive liposomes, applicable in drug delivery.
  • In the preparation of ionic polymer complex bulk material, which is optically active.
  • As a component of the tyrosinase-based biosensor, applicable in the detection of phenol molecules.

Other Notes

Double-chained surfactant used as lipid model, bilayer-forming amphiphile

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electrochemical detection of phenolic compounds using composite film of multiwall carbon nanotube/surfactant/tyrosinase on a carbon paste electrode
Hashemnia S, et al.
Journal of Solid State Electrochemistry, 16(2), 473-479 (2012)
Preparation of pH-sensitive liposomes retaining SOD mimic and their anticancer effect
Hiraka K, et al.
Colloids and Surfaces, B: Biointerfaces, 67(1), 54-58 (2008)
Tetrahedron Letters, 31, 5335-5335 (1990)
Dimethylditetradecylammonium bromide (2C 14 DAB) as a self-assembled surfactant coating for detection of protein-dye complexes by CE-LIF
Gerardi AR, et al.
Journal of Solid State Electrochemistry, 13(4), 633-638 (2009)
Layered nanoarchitecture of a fluorescent polyelectrolyte complex
Thunemann AF and Ruppelt D
Langmuir, 16(7), 3221-3226 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service