Skip to Content
Merck
All Photos(2)

Key Documents

420220

Sigma-Aldrich

Dihexadecyldimethylammonium bromide

97%

Synonym(s):

Dimethyldihexadecylammonium bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)15]2N(CH3)2(Br)
CAS Number:
Molecular Weight:
574.85
Beilstein:
4058078
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

159-162 °C (lit.)

SMILES string

[Br-].CCCCCCCCCCCCCCCC[N+](C)(C)CCCCCCCCCCCCCCCC

InChI

1S/C34H72N.BrH/c1-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-35(3,4)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-2;/h5-34H2,1-4H3;1H/q+1;/p-1

InChI key

VIXPKJNAOIWFMW-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maximilian Schmid et al.
Colloids and surfaces. B, Biointerfaces, 169, 298-304 (2018-05-25)
Investigating lipid ion pair formation is important for understanding the mechanisms of lipid-mediated drug resistance in bacteria. In this study we have used the charged amphiphiles dipalmitoylphosphatidylglycerol (DPPG) and dihexadecyldimethylammonium bromide (DHDAB), as a model to evaluate the formation of
Seong-Hwi Hong et al.
Genes, 13(7) (2022-07-28)
Renal cell carcinoma (RCC) frequently recurs or metastasizes after surgical resection. Everolimus, an mTOR inhibitor, is used as a second-line treatment, but the response of RCC to everolimus is insufficient. Metformin is an antidiabetic drug; recent reports have indicated its
Kseniia Sarieva et al.
Disease models & mechanisms, 16(11) (2023-11-03)
Maternal immune activation (MIA) increases the risks for neurodevelopmental disorders in offspring through inflammatory cytokines, including interleukin-6 (IL-6). We therefore aimed to establish a human two-dimensional (2D) in vitro neural model to investigate the effects of IL-6 exposure on neurodevelopment.
Arun Kumar et al.
Food chemistry, 199, 660-666 (2016-01-19)
Self-assemblies of equimolar double and single chain mixed ionic surfactants, with increasing numbers of carbon atoms of double chain surfactant, were analyzed on the basis of fluorescence and conductivity results. Attempts were also made to enhance the solubilization of curcumin
Carolina Rodriguez-Tirado et al.
Cancer research, 82(12), 2313-2326 (2022-04-27)
Cancer cells can disseminate during very early and sometimes asymptomatic stages of tumor progression. Though biological barriers to tumorigenesis have been identified and characterized, the mechanisms that limit early dissemination remain largely unknown. We report here that the orphan nuclear

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service