Skip to Content
Merck
All Photos(1)

Key Documents

G5635

Sigma-Aldrich

β-Galactosidase from Escherichia coli

Grade VIII, lyophilized powder, ≥500 units/mg protein

Synonym(s):

β-D-Galactoside galactohydrolase, Lactase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

type

Grade VIII

Quality Level

form

lyophilized powder

specific activity

≥500 units/mg protein

mol wt

465 kDa

does not contain

BSA as extender

composition

Protein, ≥60% biuret

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

β-Galactosidase is a tetramer consisting of four equal subunits of 135,000 each. It is a sulfhydryl containing enzyme, with 19 cysteine residues per subunit.

Application

β-Galactosidase is conjugated to an antibody which specifically recognizes a target molecule (enzyme immunoassay or EIA). β-Galactosidase is also used as a reporter enzyme to monitor the level of gene expression of a promoter. It may be used as a positive control protein with anti-β-galactosidase antibodies.

Biochem/physiol Actions

β-galactosidase cleaves lactose into its monosaccharide components, glucose and galactose. It also catalyses the transglycosylation of glucose into allolactose, the inducer of β-galactosidase, in a feedback loop.

Physical properties

Tetramer molecular weight 465 kDa (subunits 116.3 kDa each)

Unit Definition

One unit will hydrolyze 1.0 μmole of o-nitrophenyl β-D-galactoside to o-nitrophenol and D-galactose per min at pH 7.3 at 37 °C.

Physical form

Contains Tris buffer salts and magnesium chloride

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alan Merk et al.
IUCrJ, 7(Pt 4), 639-643 (2020-07-23)
We report the determination of the structure of Escherichia coli β-galactosidase at a resolution of ∼1.8 Å using data collected on a 200 kV CRYO ARM microscope equipped with a K3 direct electron detector. The data were collected in a single 24 h
Assays for bacterial mucin-desulfating sulfatases.
A M Roberton et al.
Methods in molecular biology (Clifton, N.J.), 125, 417-426 (2000-05-23)
Toshiya Suzuki et al.
Plant physiology, 173(1), 183-205 (2016-11-12)
Pollen exine is essential for protection from the environment of the male gametes of seed-producing plants, but its assembly and composition remain poorly understood. We previously characterized Arabidopsis (Arabidopsis thaliana) mutants with abnormal pollen exine structure and morphology that we
K Kato et al.
Journal of immunology (Baltimore, Md. : 1950), 116(6), 1554-1560 (1976-06-01)
1. A method for the conjugation of the Fab' fragment of rabbit IgG with beta-D-galactosidase from Escherichia coli is described. The method consists of two main steps: treatment of the Fab' fragments containing sulfhydryl groups with excess N,N'-o-phenylenedimaleimide, to introduce
Raimond B G Ravelli et al.
Nature communications, 11(1), 2563-2563 (2020-05-24)
The increasing demand for cryo-electron microscopy (cryo-EM) reveals drawbacks in current sample preparation protocols, such as sample waste and lack of reproducibility. Here, we present several technical developments that provide efficient sample preparation for cryo-EM studies. Pin printing substantially reduces

Articles

For use as a marker in SDS-PAGE; Albumin from chicken egg white, For use as a marker in SDS-PAGE; L-Lactic Dehydrogenase from rabbit muscle, Type XI, lyophilized powder, 600-1,200 units/mg protein

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service