Skip to Content
Merck
All Photos(1)

Key Documents

394408

Sigma-Aldrich

1-(Bromoacetyl)pyrene

97%

Synonym(s):

2-Bromo-1-(1-pyrenyl)ethanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H11BrO
CAS Number:
Molecular Weight:
323.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

mp

129-131 °C (lit.)

functional group

bromo
ketone

SMILES string

BrCC(=O)c1ccc2ccc3cccc4ccc1c2c34

InChI

1S/C18H11BrO/c19-10-16(20)14-8-6-13-5-4-11-2-1-3-12-7-9-15(14)18(13)17(11)12/h1-9H,10H2

InChI key

KAEDEGFCOPIKKM-UHFFFAOYSA-N

Related Categories

General description

1-(Bromoacetyl)pyrene (BAP) is a pyrene derivative. It has been synthesized by reacting cupric bromide with 1-acetylpyrene. Studies suggest that the introduction of a bromoacetyl chromophoric moiety to pyrene drastically increases the photoinitiating efficiency of pyrenes.

Application

1-(Bromoacetyl)pyrene is suitable for use in the following studies:
  • As an initiator in the bulk polymerization of 2-ethyl-2-oxazoline to generate pyrene labelled poly(2-ethyl-2-oxazoline) (PETOX-py).
  • As a fluorophore in the generation of podand-type fluoroionophores with two pyrene moieties.
  • As a fluorescent labeling agent for the determination of okadaic acid toxin by HPLC with fluorescence detection.
It may also be used in the following studies:
  • As a photoremovable protecting group for carboxylic acids and amino acids.
  • As a photoinitiator in the photopolymerization of styrene with methylmethacrylate.
  • As a reactant in the synthesis of potentially tetradentate pyrene appended ligands.
  • As a derivatizing agent of dialkyl phosphates (DAP) in the HPTLC method of quantitative determination of DAP in fruit juices.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Juraj Kronek et al.
Journal of materials science. Materials in medicine, 22(7), 1725-1734 (2011-05-24)
Poly(2-oxazolines) with varying alkyl chain lengths (e.g., methyl, ethyl, aryl) and molar masses have been tested for cell cytotoxicity in vitro. A standard 3-(4,5-dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT) assay was used for the estimation of cell viability. Two monomers, 2-methyl-2-oxazoline and 2-ethyl-2-oxazoline
Nicola M Cox et al.
Dalton transactions (Cambridge, England : 2003), 41(5), 1568-1573 (2011-12-07)
A new method for assessing the topology of metallosupramolecular assemblies using pyrene-appended ligands is reported. Two potentially tetradentate ligands containing one (L(1)) and two (L(2)) terminal pyrene moieties were synthesised and their complexes with Cu(+) and Cd(2+) were characterised. Photophysical
S S Kelly et al.
Journal of chromatography. A, 749(1-2), 33-40 (1996-10-18)
The rare diarrhetic shellfish toxin, dinophysistoxin-2 (DTX-2), was isolated from the digestive glands of mussels (Mytilus edulis). This was achieved by chromatography on silica and Sephadex LH-20 followed by reversed-phase solid phase extraction and semi-preparative high-performance liquid chromatography (HPLC) with
Copolymerization of n-butylacrylate with styrene by a novel photoinitiator, 1-(bromoacetyl) pyrene.
Mishra A and Daswal S
Journal of Applied Polymer Science, 102.4, 3233-3239 (2006)
H Ochiai et al.
Journal of chromatography. B, Biomedical sciences and applications, 694(1), 211-217 (1997-06-20)
By using a fluorescent derivatization and column-switching technique, a highly sensitive and selective high-performance liquid chromatographic (HPLC) method has been developed for the determination of simvastatin (I, beta-hydroxy-delta-lactone form) and its active hydrolyzed metabolite (II, beta,delta-dihydroxy acid form of I)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service