Skip to Content
Merck
All Photos(1)

Key Documents

761885

Sigma-Aldrich

1-(Bromomethyl)pyrene

Synonym(s):

1-Pyrenylmethyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H11Br
CAS Number:
Molecular Weight:
295.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

solid

mp

140-145 °C

functional group

bromo

SMILES string

BrCc1ccc2ccc3cccc4ccc1c2c34

InChI

1S/C17H11Br/c18-10-14-7-6-13-5-4-11-2-1-3-12-8-9-15(14)17(13)16(11)12/h1-9H,10H2

InChI key

UGMXRPVWWWDPFC-UHFFFAOYSA-N

Related Categories

Application

1-(Bromomethyl)pyrene is widely used in the synthesis of fluorophores for the fluorescent sensing of variety of analytes including metal ions like Cd2+, Zn2+ and adenosine triphosphate (ATP) sensing at physiological pH. It is also used in the synthesis of photoinitiators (PIs) for the radical polymerization of acrylates and the cationic polymerization of epoxy-silicone and vinyl ethers.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

New insights into radical and cationic polymerizations upon visible light exposure: role of novel photoinitiator systems based on the pyrene chromophore.
Tehfe M A, et al.
Polym. Chem., 4(5), 1625-1634 (2013)
A tong-like fluorescence sensor for metal ions: perfect conformational switch of hinge sugar by pyrene stacking.
Yuasa H, et al.
Organic & Biomolecular Chemistry, 2(24), 3548-3556 (2004)
A pyrenyl-appended triazole-based calix [4] arene as a fluorescent sensor for Cd2+ and Zn2+.
Park S Y, et al.
The Journal of Organic Chemistry, 73(21), 8212-8218 (2008)
Unique sandwich stacking of pyrene-adenine-pyrene for selective and ratiometric fluorescent sensing of ATP at physiological pH.
Xu Z, et al.
Journal of the American Chemical Society, 131(42), 15528-15533 (2009)
Jing Wang et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 228, 117725-117725 (2019-11-14)
We developed PIM, a pyrene-based fluorescence sensor bearing an imidazole moiety and a carbonyl group as the binding sites for Fe3+ ions. The pyrene-based control compounds 1 and 2 were synthesized to demonstrate the structure-activity relationships. Compound 1, which contained

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service