Skip to Content
Merck
All Photos(2)

Key Documents

372889

Sigma-Aldrich

1,3-Diethyl-1,3-diphenylurea

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[C6H5N(C2H5)]2CO
CAS Number:
Molecular Weight:
268.35
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

9.3 (vs air)

Quality Level

Assay

99%

form

solid

mp

73-75 °C (lit.)

solubility

acetone: soluble(lit.)
benzene: soluble(lit.)
ethanol: soluble(lit.)
water: insoluble(lit.)

functional group

amine

SMILES string

CCN(C(=O)N(CC)c1ccccc1)c2ccccc2

InChI

1S/C17H20N2O/c1-3-18(15-11-7-5-8-12-15)17(20)19(4-2)16-13-9-6-10-14-16/h5-14H,3-4H2,1-2H3

InChI key

PZIMIYVOZBTARW-UHFFFAOYSA-N

Gene Information

human ... EPHX2(2053)
mouse ... Ephx2(13850)

General description

1,3-Diethyl-1,3-diphenylurea is a symmetrical derivative of urea.1 1,3-Diethyl-1,3-diphenylurea is a commonly employed as stabilizer in double-base propellants. Molecule of 1,3-diethyl-1,3-diphenylurea exhibits non-crystallographic C2 symmetry. Photochemically induced fluorescence (PIF) studies of 1,3-diethyl-1,3-diphenylurea as stabilizer has been reported . 1,3-Diethyl-l,3-diphenylurea (Ethylcentralite or EC) has been reported as propellant stabilizer degradation derivative. Effect of ethyl centralite on a cyclotrimethylenetrinitramine (RDX)-based polymer-bonded explosive (PBX) has been reported.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

302.0 °F - closed cup

Flash Point(C)

150 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemical Desensitization of Explosives. Part 1. Effect of Ethyl Centralite on the Properties of an RDX-Based PBX.
Padfield J, et al.
Journal of Energetic Materials, 32(2), 106-116 (2014)
Richard Betz et al.
Acta crystallographica. Section E, Structure reports online, 67(Pt 4), o827-o827 (2011-07-15)
The mol-ecule of the title compound, C(17)H(20)N(2)O, a symmetrical derivative of urea, shows non-crystallographic C(2) symmetry. Inter-action with the aromatic system of the phenyl substituents as well as amide-type resonance is responsible for the marked planarization of the coordination environments
Synthetic Procedures Yielding Targeted Nitro and Nitroso Derivatives of the Propellant Stabilisers Diphenylamine, N-Methyl-4-nitroaniline, and N, N'-Diethyl-N, N'-diphenylurea.
Elliot MS, et al.
Propellants, Explosives, Pyrotechnics, 25(1), 31-36 (2000)
Colby E Ott et al.
Journal of forensic sciences, 65(6), 1935-1944 (2020-08-26)
The increasing demand for rapid methods to identify both inorganic and organic gunshot residues (IGSR and OGSR) makes electrochemical methods, an attractive screening tool to modernize current practice. Our research group has previously demonstrated that electrochemical screening of GSR samples
Alireza Farokhcheh et al.
Forensic science international, 240, 35-40 (2014-05-06)
The photochemically induced fluorescence (PIF) studies of 1,3-diethyl-1,3-diphenylurea or ethyl centralite (EC) as stabilizer have been carried out under various conditions. Influences of solvent type (water, methanol, acetonitrile and chloroform), oxygen dependence and UV irradiation time on the spectroscopic properties

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service