Skip to Content
Merck
All Photos(3)

Key Documents

524980

Sigma-Aldrich

Dibutyl phthalate

99%

Synonym(s):

n-Butyl phthalate, DBP, Phthalic acid dibutyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4-1,2-[CO2(CH2)3CH3]2
CAS Number:
Molecular Weight:
278.34
Beilstein:
1914064
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
bp:
340 °C (lit.)
vapor pressure:
1 mmHg ( 147 °C)

vapor density

9.6 (vs air)

Quality Level

vapor pressure

1 mmHg ( 147 °C)

Assay

99%

form

liquid

autoignition temp.

756 °F

expl. lim.

0.47 %, 236 °F

impurities

≤0.005% Acidity (as Phthalic acid)
≤0.15% water

color

APHA: ≤20

refractive index

n20/D 1.492 (lit.)

pH

7 (20 °C, 10 mg/L)

bp

340 °C (lit.)

mp

−35 °C (lit.)

density

1.043 g/mL at 25 °C (lit.)

functional group

ester

SMILES string

CCCCOC(=O)c1ccccc1C(=O)OCCCC

InChI

1S/C16H22O4/c1-3-5-11-19-15(17)13-9-7-8-10-14(13)16(18)20-12-6-4-2/h7-10H,3-6,11-12H2,1-2H3

InChI key

DOIRQSBPFJWKBE-UHFFFAOYSA-N

Gene Information

mouse ... Esr1(13982)

Looking for similar products? Visit Product Comparison Guide

General description

Dibutyl phthalate (DBP) is most commonly used as a plasticizer in a variety of household products. It has the ability to leach and evaporate into the environment while using or disposing of these products. Therefore, studies are being conducted to assess its toxic and apoptotic effects. Studies have also indicated that exposure to high doses of DBP may interfere with progesterone and estradiol production.

Application

Plasticizer

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

367.7 °F - open cup

Flash Point(C)

186.5 °C - open cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dibutyl phthalate (DBP)-induced apoptosis and neurotoxicity are mediated via the aryl hydrocarbon receptor (AhR) but not by estrogen receptor alpha (ER?), estrogen receptor beta (ER?), or peroxisome proliferator-activated receptor gamma (PPAR?) in mouse cortical neurons.
Wojtowicz A
Neurotoxicity Research, 31(1), 77-89 (2017)
Dibutyl phthalate impairs steroidogenesis and a subset of LH-dependent genes in cultured human mural granulosa cell in vitro.
Adir M
Reproductive Toxicology, 69, 13-18 (2017)
Linda V Sinclair et al.
Immunometabolism, 2(4), e200029-e200029 (2020-09-04)
Assays to monitor the metabolic state or nutrient uptake capacity of immune cells at a single cell level are increasingly in demand. One assay, used by many immunologists, employs 2-(N-(7-Nitrobenz-2-oxa-1,3-diazol-4-yl)Amino)-2-Deoxyglucose (2-NBDG), a fluorescent analogue of 2-deoxyglucose (2DG), as a substrate
Damien Hunter et al.
International journal of molecular sciences, 22(8) (2021-05-01)
Xenobiotic exposure during pregnancy and lactation has been linked to perinatal changes in male reproductive outcomes and other endocrine parameters. This pilot study wished to assess whether brief maternal exposure of rats to xenobiotics dibutyl phthalate (DBP) or diethylstilbestrol (DES)
Harpreet Bhatia et al.
Aquatic toxicology (Amsterdam, Netherlands), 149, 103-115 (2014-03-01)
Phthalic acid esters (PAEs) are a class of synthetic industrial chemicals commonly found in the aquatic environment worldwide. PAEs have been recognised as anti-androgens in male mammals but little is known about their endocrine disrupting effects in fish. This study

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service