Skip to Content
Merck
All Photos(2)

Key Documents

252441

Sigma-Aldrich

Rhodamine 6G perchlorate

Dye content 99 %

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H30N2O3 · HClO4
CAS Number:
Molecular Weight:
543.01
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

form

solid

composition

Dye content, 99%

mp

263-265 °C (lit.)

λmax

528 nm

SMILES string

OCl(=O)(=O)=O.CCNc1cc2OC3=C\C(=N/CC)C(C)=CC3=C(c2cc1C)c4ccccc4C(=O)OCC

InChI

1S/C28H30N2O3.ClHO4/c1-6-29-23-15-25-21(13-17(23)4)27(19-11-9-10-12-20(19)28(31)32-8-3)22-14-18(5)24(30-7-2)16-26(22)33-25;2-1(3,4)5/h9-16,29H,6-8H2,1-5H3;(H,2,3,4,5)/b30-24+;

InChI key

HDAFVOZRAUFNQH-WTKGSRSZSA-N

General description

Rhodamine 6G perchlorate is an amphiphilic organic dye that can have a high quantum yield and a tunability in the range of 550-600 nm. It is used in dye lasers and can also be used in the textile industry.[1]

Application

Rhodamine 6G perchlorate is a xanthene derivative that can be incorporated with nanomaterials to form a nanocluster, which can be used in biological sensing and imaging applications. It may also be used in the development of solid state laser dye for optics and remote sensing.[2][3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

1H and 13C NMR spectra of commercial rhodamine ester derivatives.
Ramos SS, et al.
Magnetic Resonance in Chemistry, 38(6), 475-478 (2000)
Dye-capped gold nanoclusters: photoinduced morphological changes in gold/rhodamine 6G nanoassemblies.
Chandrasekharan N, et al.
The Journal of Physical Chemistry B, 104(47), 11103-11109 (2000)
Rhodamine 6G impregnated porous silica: A photoluminescence study.
Anedda A, et al.
Materials Science and Engineering, C, 25(5-8), 641-644 (2005)
Jing Chen et al.
Biosensors & bioelectronics, 44, 191-197 (2013-02-23)
Nanoscale gaps in noble metal films can produce intense electromagnetic enhancement. When Raman-active molecules are positioned in these regions, their surface-enhanced Raman scattering (SERS) signals can be dramatically enhanced. However, the lack of convenient and reliable fabrication methods with ultrasmall
Xinhua Huang et al.
Langmuir : the ACS journal of surfaces and colloids, 29(10), 3223-3233 (2013-02-22)
Tetraarylmethane compounds consisting of two pyrogallol and two aniline units, namely, Ar2CAr'2 {Ar = 3,4,5-C6H2(OH)3 and Ar' = 3,5-R2-4-C6H2NH2 [R = Me (1), iPr (2)]} exhibit excellent self-assembly behavior. Compound 1 yields size-tunable hollow nanospheres (HNSs) with a narrow size

Articles

Developed in the last several years, fluorescence quenching microscopy (FQM) has enabled rapid, inexpensive, and high-fidelity visualization of two-dimensional (2D) materials such as graphene-based sheets and MoS2.

Developed in the last several years, fluorescence quenching microscopy (FQM) has enabled rapid, inexpensive, and high-fidelity visualization of two-dimensional (2D) materials such as graphene-based sheets and MoS2.

Fluorescence quenching microscopy visualizes 2D materials like graphene and MoS2 rapidly, inexpensively, and with high fidelity.

Fluorescence quenching microscopy visualizes 2D materials like graphene and MoS2 rapidly, inexpensively, and with high fidelity.

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service