Skip to Content
Merck
All Photos(1)

Key Documents

366404

Sigma-Aldrich

1,1,4,7,10,10-Hexamethyltriethylenetetramine

97%

Synonym(s):

HMTETA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2NCH2CH2N(CH3)CH2-]2
CAS Number:
Molecular Weight:
230.39
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.456 (lit.)

bp

130 °C/11 mmHg (lit.)

density

0.847 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

CN(C)CCN(C)CCN(C)CCN(C)C

InChI

1S/C12H30N4/c1-13(2)7-9-15(5)11-12-16(6)10-8-14(3)4/h7-12H2,1-6H3

InChI key

DWFKOMDBEKIATP-UHFFFAOYSA-N

General description

1,1,4,7,10,10-Hexamethyltriethylenetetramine is a polyamines additive, has been reported as an efficient reagent for the problematic Koenigs-Knorr glucuronidation.

Application

1,1,4,7,10,10-Hexamethyltriethylenetetramine may be used as reagent in the synthesis of ideal linear random copolymers containing both vinyl polymer and polyester units in a single polymer chain. 1,1,4,7,10,10-Hexamethyltriethylenetetramine complexed with CuBr constitutes catalytic complex, used in the copolymerization of poly[ε-caprolactone] with N,N-dimethylamino-2-ethyl methacrylate monomers by atom-transfer radical polymerization (ATRP). It may be used as catalyst in the aqueous surface-initiated-ATRP to grow poly(N,N-dimethylacrylamide) (PDMA).

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

215.6 °F - closed cup

Flash Point(C)

102 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bryan R Coad et al.
Langmuir : the ACS journal of surfaces and colloids, 23(23), 11791-11803 (2007-10-11)
We report the use of aqueous surface-initiated atom transfer radical polymerization (SI-ATRP) to grow polymer brushes from a "gigaporous" polymeric chromatography support for use as a novel size exclusion chromatography medium. Poly(N,N-dimethylacrylamide) (PDMA) was grown from hydrolyzable surface initiators via
Stefanie L Baker et al.
Nature communications, 10(1), 4718-4718 (2019-10-19)
Almost all commercial proteins are purified using ammonium sulfate precipitation. Protein-polymer conjugates are synthesized from pure starting materials, and the struggle to separate conjugates from polymer, native protein, and from isomers has vexed scientists for decades. We have discovered that
Laetitia Mespouille et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 14(21), 6369-6378 (2008-06-10)
Adaptive and amphiphilic poly(N,N-dimethylamino-2-ethyl methacrylate-graft-poly[epsilon-caprolactone]) co-networks (netP(DMAEMA-g-PCL)) were synthesized from a combination of controlled polymerization techniques. Firstly, PCL cross-linkers were produced by ring-opening polymerization (ROP) of epsilon-CL initiated by 1,4-butane-diol and catalyzed by tin(II) 2-ethylhexanoate ([Sn(Oct)2]), followed by the quantitative
Weihang Ji et al.
Biomacromolecules, 18(8), 2583-2593 (2017-06-29)
Antibacterial polymers are potentially powerful biocides that can destroy bacteria on contact. Debate in the literature has surrounded the mechanism of action of polymeric biocides and the propensity for bacteria to develop resistance to them. There has been particular interest
Masato Mizutani et al.
Journal of the American Chemical Society, 132(21), 7498-7507 (2010-05-12)
All polymerization reactions are categorized into two large different families, chain- and step-growth polymerizations, which are typically incompatible. Here, we report the simultaneous chain- and step-growth polymerization via the metal-catalyzed radical copolymerization of conjugated vinyl monomers and designed monomers possessing

Articles

Micro review of reversible addition/fragmentation chain transfer (RAFT) polymerization.

Micro review of reversible addition/fragmentation chain transfer (RAFT) polymerization.

Micro review of reversible addition/fragmentation chain transfer (RAFT) polymerization.

Micro review of reversible addition/fragmentation chain transfer (RAFT) polymerization.

Protocols

We presents an article featuring procedures that describe polymerization of methyl methacrylate and vinyl acetate homopolymers and a block copolymer as performed by researchers at CSIRO.

We present an article about RAFT, or Reversible Addition/Fragmentation Chain Transfer, which is a form of living radical polymerization.

Polymerization via ATRP procedures demonstrated by Prof. Dave Haddleton's research group at the University of Warwick.

Polymerization via ATRP procedures demonstrated by Prof. Dave Haddleton's research group at the University of Warwick.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service