Skip to Content
Merck
All Photos(3)

Key Documents

254185

Sigma-Aldrich

Copper(I) bromide

99.999% trace metals basis

Synonym(s):

Cuprous bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CuBr
CAS Number:
Molecular Weight:
143.45
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
powder

grade

for analytical purposes

Assay

99.999% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: copper

impurities

≤15.0 ppm Trace Metal Analysis

mp

504 °C (lit.)

density

4.71 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

[Cu]Br

InChI

1S/BrH.Cu/h1H;/q;+1/p-1

InChI key

NKNDPYCGAZPOFS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Copper(I) bromide is a faint green inorganic diamagnetic solid that has a polymeric structure like zinc sulfide. It is insoluble in water and widely used in the field of catalysis, nanomaterial synthesis, and solar cells.

Application

Copper(I) bromide can be used:
  • As a solution-processable hole transport layer (HTL) for organic solar cells (OSCs). CuBr enhances the power conversion efficiency of solar cells.
  • As a precursor to synthesize photoluminescent metal organic frameworks(MOFs).
  • To synthesize highly emissive three-coordinate copper(I) complexes as a dopant in OLEDs with maximum external quantum efficiency of 21.3%.
  • As a catalyst with ligands for atom transfer radical polymerization (ATRP) reaction of tertiary alkyl halides in 14 different solvents to determine the rate constant.
  • To synthesize macrocyclic polymer via intramolecular radical trap-assisted atom transfer radical coupling.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An acs-Type Metal--Organic Framework with an Unprecedented Undecanuclear Copper Cluster as Secondary Building Unit
Sheng Hu, et al.
European Journal of Inorganic Chemistry, 2012, 3669-3673 (2012)
Highly Efficient Green Organic Light-Emitting Diodes Containing Luminescent Three-Coordinate Copper(I) Complexes
Masashi Hashimoto, et al.
Journal of the American Chemical Society, 133, 10348-10351 (2011)
Copper Bromide as an Efficient Solution-Processable Hole Transport Layer for Organic Solar Cells: Effect of Solvents
Ranoo Bhargav, et al.
ACS Omega, 4, 6028-6034 (2019)
Carla Sardo et al.
International journal of pharmaceutics, 563, 347-357 (2019-04-03)
Sustained pulmonary delivery of tobramycin from microparticles composed of drug/polymer nanocomplexes offers several advantages against traditional delivery methods. Namely, in patients with cystic fibrosis, microparticle delivery can protect the tobramycin being delivered from strong mucoadhesive interactions, thus avoiding effects on
Denissova I and Barriault L et al.
Handbook of Reagents for Organic Synthesis null

Articles

Micro review of reversible addition/fragmentation chain transfer (RAFT) polymerization.

Micro review of reversible addition/fragmentation chain transfer (RAFT) polymerization.

Micro review of reversible addition/fragmentation chain transfer (RAFT) polymerization.

Micro review of reversible addition/fragmentation chain transfer (RAFT) polymerization.

Protocols

We presents an article featuring procedures that describe polymerization of methyl methacrylate and vinyl acetate homopolymers and a block copolymer as performed by researchers at CSIRO.

We present an article about RAFT, or Reversible Addition/Fragmentation Chain Transfer, which is a form of living radical polymerization.

Polymerization via ATRP procedures demonstrated by Prof. Dave Haddleton's research group at the University of Warwick.

Polymerization via ATRP procedures demonstrated by Prof. Dave Haddleton's research group at the University of Warwick.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service