Skip to Content
Merck
All Photos(3)

Documents

327395

Sigma-Aldrich

Germanium

powder, −100 mesh, ≥99.999% trace metals basis

Synonym(s):

Elemental germanium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ge
CAS Number:
Molecular Weight:
72.64
EC Number:
MDL number:
UNSPSC Code:
12141716
PubChem Substance ID:
NACRES:
NA.23

Assay

≥99.999% trace metals basis

form

powder

resistivity

53 Ω-cm, 20°C

particle size

−100 mesh

bp

2830 °C (lit.)

mp

937 °C (lit.)

density

5.35 g/mL at 25 °C (lit.)

SMILES string

[Ge]

InChI

1S/Ge

InChI key

GNPVGFCGXDBREM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

The product can serve as one of the precursors for the formation of highly porous ZrO2:Tb3+ nanophosphor with excellent tunable photoluminescence and photocatalytic activities.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Flam. Sol. 1 - Repr. 2 - STOT RE 2 Oral

Target Organs

Kidney

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thermoluminescence glow curve for UV induced ZrO2:Ti phosphor with variable concentration of dopant and various heating rate
Tiwari N, et al.
Journal of Radiation Research and Applied Sciences, 7(4), 542-549 (2014)
Phase transformation of ZrO 2: Tb 3+ nanophosphor: Color tunable photoluminescence and photocatalytic activities.
Vidya YS, et al.
J. Alloy Compounds, 622, 86-96 (2015)
Michael Oehme et al.
Optics express, 21(2), 2206-2211 (2013-02-08)
In this paper we investigate the influence of n-type doping in Ge light emitting diodes on Si substrates on the room temperature emission spectrum. The layer structures are grown with a special low temperature molecular beam epitaxy process resulting in
Jin Liu et al.
Dalton transactions (Cambridge, England : 2003), 42(14), 5092-5099 (2013-02-13)
In this study, Zn2GeO4 hollow spheres were successfully fabricated by a template-engaged approach using zinc hydroxide carbonate (Zn4CO3(OH)6·H2O, ZHC) spheres as the template. During the hydrothermal process, Zn(2+) dissolved from the surface of the ZHC spheres could rapidly react with
Lu Dai et al.
Nanoscale, 5(3), 971-976 (2012-12-15)
The controllable fabrication of self-scrolling SiGe/Si/Cr helical nanoribbons on Si(111) substrates is investigated. The initial lateral etching profile of the Si(111) substrates shows a 2-fold rotational symmetry using 4% ammonia solution, which provides guidance for initial scrolling of one-end-fixed nanoribbons

Articles

Higher transition metal silicides are ideal for anisotropic thermoelectric conversion due to their Seebeck coefficient anisotropy and mechanical properties.

Higher transition metal silicides are ideal for anisotropic thermoelectric conversion due to their Seebeck coefficient anisotropy and mechanical properties.

Higher transition metal silicides are ideal for anisotropic thermoelectric conversion due to their Seebeck coefficient anisotropy and mechanical properties.

Higher transition metal silicides are ideal for anisotropic thermoelectric conversion due to their Seebeck coefficient anisotropy and mechanical properties.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service