Skip to Content
Merck
All Photos(1)

Documents

267961

Sigma-Aldrich

Arsenic

powder, ≥99.997% trace metals basis

Synonym(s):

Arsenic black, Arsenic-75

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
As
CAS Number:
Molecular Weight:
74.92
EC Number:
MDL number:
UNSPSC Code:
12141906
PubChem Substance ID:
NACRES:
NA.23

Assay

≥99.997% trace metals basis

form

powder

resistivity

33.3 μΩ-cm

bp

613 °C (lit.)

mp

817 °C (lit.)

density

5.727 g/mL at 25 °C (lit.)

SMILES string

[As]

InChI

1S/As

InChI key

RQNWIZPPADIBDY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A - Repr. 1A - STOT RE 1 Oral

Target Organs

Respiratory organs,Cardio-vascular system,Gastro-intestinal system,Central nervous system,Nervous system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arsenic binding to proteins.
Shengwen Shen et al.
Chemical reviews, 113(10), 7769-7792 (2013-07-03)
Meryene C Teixeira et al.
Food chemistry, 154, 38-43 (2014-02-13)
We have developed an eletroanalytical method that employs Cu(2+) solutions to determine arsenic in sugarcane brandy using an electrode consisting of carbon paste modified with carbon nanotubes (CNTPE) and polymeric resins. We used linear sweep (LSV) and differential-pulse (DPV) voltammetry
Ilona Dudka et al.
Environment international, 68, 71-81 (2014-04-10)
Environmental metabonomics is the application of metabonomics to characterize the interactions of organisms with their environment. Metabolic profiling is an exciting addition to the armory of the epidemiologist for the discovery of new disease risk biomarkers and diagnostics. This work
Samuel M Cohen et al.
Critical reviews in toxicology, 43(9), 711-752 (2013-09-18)
Inorganic arsenic (iAs) at high exposures is a human carcinogen, affecting mainly the urinary bladder, lung and skin. We present an assessment of the mode of action (MOA) of iAs's carcinogenicity based on the United States Environmental Protection Agency/International Programme
Luis Rodríguez-Lado et al.
Science (New York, N.Y.), 341(6148), 866-868 (2013-08-24)
Arsenic-contaminated groundwater used for drinking in China is a health threat that was first recognized in the 1960s. However, because of the sheer size of the country, millions of groundwater wells remain to be tested in order to determine the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service