Skip to Content
Merck
All Photos(2)

Documents

302155

Sigma-Aldrich

(S)-(+)-1-Phenyl-1,2-ethanediol

99%

Synonym(s):

(+)-Styrene glycol, (S)-(+)-Phenylethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CH(C6H5)OH
CAS Number:
Molecular Weight:
138.16
Beilstein:
3196197
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

optical activity

[α]18/D +66°, c = 1 in chloroform

mp

64-67 °C (lit.)

SMILES string

OC[C@@H](O)c1ccccc1

InChI

1S/C8H10O2/c9-6-8(10)7-4-2-1-3-5-7/h1-5,8-10H,6H2/t8-/m1/s1

InChI key

PWMWNFMRSKOCEY-MRVPVSSYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

320.0 °F - closed cup

Flash Point(C)

160.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rongzhen Zhang et al.
Wei sheng wu xue bao = Acta microbiologica Sinica, 49(2), 204-209 (2009-05-19)
To prepare (S)-1-phenyl-1,2-ethanediol by a one-step method, we constructed an enzyme coupled system consisting of (R)- and (S)-specific carbonyl reductases in Escherichia coli. The genes coding (R)- and (S)-specific carbonyl reductases from Candida parapsilosis were inserted into a co-expression vector
Li Cao et al.
Biotechnology and bioengineering, 94(3), 522-529 (2006-02-25)
Soluble epoxide hydrolase (EH) from the potato Solanum tuberosum and an evolved EH of the bacterium Agrobacterium radiobacter AD1, EchA-I219F, were purified for the enantioconvergent hydrolysis of racemic styrene oxide into the single product (R)-1-phenyl-1,2-ethanediol, which is an important intermediate
Jin-Zhao Wang et al.
Biomedical chromatography : BMC, 21(5), 497-501 (2007-03-16)
A simple HPLC method for the simultaneous determination of phenylglyoxylic acid (PGA), mandelic acid (MA), styrene glycol (SG) and hippuric acid (HA) in cell culture medium was developed. Analysis was performed on a C(18) column with a mobile phase composed
Rongzhen Zhang et al.
Applied biochemistry and biotechnology, 160(3), 868-878 (2009-02-12)
An R-specific carbonyl reductase from Candida parapsilosis (CprCR) catalyzes the transformation of (R)-1-phenyl-1,2-ethanediol from 2-hydroxyacetophenone. The gene rcr coding CprCR contains a few codons rarely used by Escherichia coli. In order to improve chiral alcohol production, three codon variants Delta24
Yao Nie et al.
Organic & biomolecular chemistry, 9(11), 4070-4078 (2011-04-21)
The application of biocatalysis to the synthesis of chiral molecules is one of the greenest technologies for the replacement of chemical routes due to its environmentally benign reaction conditions and unparalleled chemo-, regio- and stereoselectivities. We have been interested in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service