
(S)-(+)-1-Phenyl-1,2-ethanediol

Synonym(s):
(+)-Styrene glycol, (S)-(+)-Phenylethylene glycol
Linear Formula:
HOCH2CH(C6H5)OH
CAS Number:
Molecular Weight:
138.16
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing