Skip to Content
Merck
All Photos(2)

Key Documents

289337

Sigma-Aldrich

Cesium nitrate

99%

Synonym(s):

Nitric acid cesium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CsNO3
CAS Number:
Molecular Weight:
194.91
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
crystals and lumps

Quality Level

Assay

99%

form

crystals and lumps

mp

414 °C (lit.)

density

3.685 g/mL at 25 °C (lit.)

SMILES string

[Cs+].[O-]N(=O)=O

InChI

1S/Cs.NO3/c;2-1(3)4/q+1;-1

InChI key

NLSCHDZTHVNDCP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Cesium nitrate is used in the synthesis of cesium nitratocuprate, Cs2[Cu(NO3)4], which crystallizes in a square-planar coordination around the Cu center. CsNO3 may also be used in the preparation of a low temperature CO conversion Cu-Zn-Al catalysts.
Used in the synthesis of cesium nitratocuprate, Cs2[Cu(NO3)4], which crystallizes in a square-planar coordination around the Cu center.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Ox. Sol. 1 - Repr. 2

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Morozov, I.V. et al.
Zeitschrift fur Anorganische und Allgemeine Chemie, 624, 1543-1543 (1998)
Himadri Rajput et al.
Chemosphere, 218, 687-695 (2018-12-05)
In this study, novel Cesium (Cs) doped TiO2 nanotubes photoelectrode (Cs/TiO2NTs) were synthesized by simple electrochemical anodization method and characterized by several physicochemical techniques. In particular, the photocatalytic (PC), electrocatalytic (EC) and photoelectrocatalytic (PEC) activity of newly synthesized Cs/TiO2NTs electrodes
Delhia Alby et al.
Journal of hazardous materials, 368, 661-669 (2019-02-08)
An alternative microwave-assisted hydrothermal route for the preparation of manganate nanoflowers under basic conditions has been proposed in view of potential uses in selective retention of strontium from multicomponent aqueous streams. Based on the combination of such characterization techniques as
Morozov, I.V. et al.
Zeitschrift fur Anorganische und Allgemeine Chemie, 624, 1543-1543 (1998)

Articles

Colloidal quantum dots (CQDs) are semiconducting crystals of only a few nanometers (ca. 2–12 nm) coated with ligand/surfactant molecules to help prevent agglomeration.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service