Skip to Content
Merck
All Photos(1)

Key Documents

83750

Sigma-Aldrich

Rhodium(III) nitrate hydrate

~36% rhodium (Rh) basis

Synonym(s):

Rhodium trinitrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
N3O9Rh · xH2O
CAS Number:
Molecular Weight:
288.92 (anhydrous basis)
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
powder

grade

for analytical purposes

form

powder

concentration

~36% Rh

SMILES string

O.[Rh+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/3NO3.H2O.Rh/c3*2-1(3)4;;/h;;;1H2;/q3*-1;;+3

InChI key

QKLCKVVKVHCMIC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Rhodium(III) nitrate hydrate is a crystallinecompound widely used in the field of catalysis, electro-plating, and glassindustry

Application

Rhodium(III) nitrate hydrate can be used as a precursor:
  • To synthesize Rh-doped catalysts for various chemical transformations.
  • With iron(III) nitrate nonahydrate to synthesis Fe–Rh/TiO2 catalysts for hydrogenation reaction.
  • For Ethene Hydroformylation, a bimetallic Rh/Co catalyst is synthesized by combining Rhodium(III) nitrate hydrate with Co(NO3)2 .6H2O.
  • For the simultaneous electrodeposition of Cu and Rh from an aqueous nitrate solution.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Flame over circleCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Ox. Sol. 3 - Skin Corr. 1B

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The effect of Fe?Rh alloying on CO hydrogenation to C2+ oxygenates
Robert M. Palomino, et al.
Chinese Journal of Catalysis, 329, 87-94 (2015)
Insights into the Bimetallic Effects of a RhCo Catalyst for Ethene Hydroformylation: Experimental and DFT Investigations
Ning Huang, et al.
Industrial & Engineering Chemistry Research, 59, Ning Huang-Ning Huang (2020)
A study of Cu-Rh electrodeposition
Dimitra Anastasiadou, et al.
ChemElectroChem, 10, e202200842-e202200842 (2023)
The role of rhodium in the mechanism of the water-gas shift over zirconia supported iron oxide
Hakeem AA, et al.
J. Catal., 313, 34-45 (2014)
Effect of rhodium on the water-gas shift performance of Fe2O3/ZrO2 and CeO2/ZrO2: Influence of rhodium precursor.
Hakeem AA,et al.
Catalysis Today, 242, 168-177 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service