Skip to Content
Merck
All Photos(7)

Documents

202126

Sigma-Aldrich

Cesium carbonate

99.9% trace metals basis

Synonym(s):

Carbonic acid dicesium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cs2CO3
CAS Number:
Molecular Weight:
325.82
Beilstein:
4546405
EC Number:
MDL number:
UNSPSC Code:
12352301
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99.9% trace metals basis

mp

610 °C (dec.) (lit.)

SMILES string

[Cs+].[Cs+].[O-]C([O-])=O

InChI

1S/CH2O3.2Cs/c2-1(3)4;;/h(H2,2,3,4);;/q;2*+1/p-2

InChI key

FJDQFPXHSGXQBY-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Catalyst for:
Aerobic oxidation of primary alcohols
1,4-addition reactions

Reagent for:
Synthesis of phosphazene derivatives
Constrictive binding interactions
Intramolecular C-N cross coupling reactions
Dehydrohalogenative polycondensation reactions
Promotes the efficient O-alkylation of alcohols to form mixed alkyl carbonates.

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Repr. 2 - STOT RE 2 Oral

Target Organs

Kidney,Adrenal gland,Testes

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael A Becker et al.
Nature, 553(7687), 189-193 (2018-01-13)
Nanostructured semiconductors emit light from electronic states known as excitons. For organic materials, Hund's rules state that the lowest-energy exciton is a poorly emitting triplet state. For inorganic semiconductors, similar rules predict an analogue of this triplet state known as
Ming Liu et al.
Advanced science (Weinheim, Baden-Wurttemberg, Germany), 4(11), 1700335-1700335 (2017-12-05)
Bright and stable blue emitters with narrow full-width at half-maxima are particularly desirable for applications in television displays and related technologies. Here, this study shows that doping aluminum (Al
Chengxi Zhang et al.
Beilstein journal of nanotechnology, 8, 2521-2529 (2017-12-21)
Colloidal perovskite nanocrystals comprised of all inorganic cesium lead halide (CsPbX
Daniel Amgar et al.
Nano letters, 17(2), 1007-1013 (2017-01-18)
Perovskite nanostructures, both hybrid organo-metal and fully inorganic perovskites, have gained a lot of interest in the past few years for their intriguing optical properties in the visible region. We report on inorganic cesium lead bromide (CsPbBr
Erin M Sanehira et al.
Science advances, 3(10), eaao4204-eaao4204 (2017-11-04)
We developed lead halide perovskite quantum dot (QD) films with tuned surface chemistry based on A-site cation halide salt (AX) treatments. QD perovskites offer colloidal synthesis and processing using industrially friendly solvents, which decouples grain growth from film deposition, and

Articles

Preformed catalysts in the kit are stable but become air-sensitive when activated; Schlenk technique aids scale-up.

Preformed catalysts in the kit are stable but become air-sensitive when activated; Schlenk technique aids scale-up.

Preformed catalysts in the kit are stable but become air-sensitive when activated; Schlenk technique aids scale-up.

Preformed catalysts in the kit are stable but become air-sensitive when activated; Schlenk technique aids scale-up.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service