Skip to Content
Merck
All Photos(3)

Key Documents

271365

Sigma-Aldrich

Ethoxyacetylene solution

~40 wt. % in hexanes

Synonym(s):

1-Ethoxyethyne, Ethoxyethyne, Ethyl ethynyl ether, Ethynyl ethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OC≡CH
CAS Number:
Molecular Weight:
70.09
Beilstein:
741882
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

liquid

Quality Level

concentration

~40 wt. % in hexanes

refractive index

n20/D 1.378

density

0.732 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CCOC#C

InChI

1S/C4H6O/c1-3-5-4-2/h1H,4H2,2H3

InChI key

WMYNMYVRWWCRPS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used with N-vinylamides and triflic anhydride in a direct, three-component synthesis of pyridines. Also used to prepare α,β-unsaturated esters from ketones via a Meyer-Schuster rearrangement.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-14.8 °F

Flash Point(C)

-26 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Direct synthesis of pyridine derivatives.
Mohammad Movassaghi et al.
Journal of the American Chemical Society, 129(33), 10096-10097 (2007-08-01)
Synlett, 949-949 (2007)
Nicholas D Stebbins et al.
Biomacromolecules, 16(11), 3632-3639 (2015-10-10)
Sugar alcohols, such as mannitol and xylitol, are biocompatible polyols that have been used to make highly cross-linked polyester elastomers and dendrimers for tissue engineering and drug delivery. However, research that utilizes the secondary hydroxyl groups as sites for pendant

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service