Skip to Content
Merck
All Photos(1)

Documents

83695

Sigma-Aldrich

Rhodamine 110 chloride

suitable for fluorescence, BioReagent, ≥99.0% (UV)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H14N2O3 · HCl
CAS Number:
Molecular Weight:
366.80
Beilstein:
4631860
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.32

product line

BioReagent

Assay

≥99.0% (UV)

form

crystals

mp

>300 °C (lit.)

solubility

ethanol: soluble
methanol: soluble

fluorescence

λex 498 nm; λem 520 nm in methanol

suitability

suitable for fluorescence

SMILES string

Cl.Nc1ccc2c(OC3=CC(=N)C=CC3=C2c4ccccc4C(O)=O)c1

InChI

1S/C20H14N2O3.ClH/c21-11-5-7-15-17(9-11)25-18-10-12(22)6-8-16(18)19(15)13-3-1-2-4-14(13)20(23)24;/h1-10,21H,22H2,(H,23,24);1H

InChI key

JNGRENQDBKMCCR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Rhodamine 110 or R110 is a triphenylmethane dye and emits a strong green-fluorescent light. The high fluorescence is attributed to the presence of amino groups as electron donating groups. Rhodamine 110 has excitation and emission maximum at 502.4/521.6 nm, respectively.

Application

Rhodamine 110 is used for the determination of protease activity with accurate kinetic parameters. It is used to measure caspase activity in cells with induced apoptosis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xian Zhang et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 59(8), 1667-1672 (2003-05-09)
A simple and sensitive fluorescence quenching method for the determination of trace nitrite has been developed. The method is based on the reaction of Rhodamine 110 with nitrite in acidic medium to form a new compound, which has much lower
Sambashiva Banala et al.
ACS chemical biology, 7(2), 289-293 (2011-10-27)
A caged rhodamine 110 derivative for the specific labeling of SNAP-tag fusion proteins is introduced. The caged rhodamine 110 derivative permits the labeling of cell surface proteins in living cells and of intracellular proteins in fixed cells. The probe requires
Aya Shibata et al.
Bioorganic & medicinal chemistry letters, 18(7), 2246-2249 (2008-03-25)
We have developed a new fluorescent probe for biological thiol. The probe was synthesized by the modification of the 2,4-dinitrobenzenesulfonyl group with rhodamine 110. The selective detection of thiol species such as cysteine or glutathione was achieved in biological conditions.
Saijie Zhu et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 37(5), 573-580 (2009-05-16)
The present study was aimed to investigate the effects of polyoxyethylene (40) stearate (PS), a non-ionic surfactant, on the activity of P-glycoprotein (P-gp) and six major cytochrome P450 (CYP) isoforms. An in vitro diffusion chamber system was utilized to estimate
H Hug et al.
Biochemistry, 38(42), 13906-13911 (1999-10-21)
Caspases (cysteine aspartate-specific proteases) are a structurally related group of cysteine proteases that cleave peptide bonds following specific recognition sequences. They play a central role in activating apoptosis of vertebrate cells. To measure apoptosis induced by various stimuli and at

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service