Skip to Content
Merck
All Photos(1)

Key Documents

244473

Sigma-Aldrich

4-Octyne

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2C≡CCH2CH2CH3
CAS Number:
Molecular Weight:
110.20
Beilstein:
1732138
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39010412
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

35 mmHg ( 37.7 °C)

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.425 (lit.)

bp

131-132 °C (lit.)

mp

−103 °C (lit.)

density

0.751 g/mL at 25 °C (lit.)

SMILES string

CCCC#CCCC

InChI

1S/C8H14/c1-3-5-7-8-6-4-2/h3-6H2,1-2H3

InChI key

GZTNBKQTTZSQNS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Octyne, also known as dipropylethyne can be used as a versatile building block in organic synthesis. The kinetics of the stereoselective semi-hydrogenation of 4-octyne (in tetrahydrofuran) was studied.

Application

4-Octyne, an electron-rich dialkylacetylene, was used in the synthesis of highly substituted 1,3-dienes.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

84.2 °F - closed cup

Flash Point(C)

29 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cyclopentadienone synthesis by rhodium (I)-catalyzed [3+ 2] cycloaddition reactions of cyclopropenones and alkynes
PA Wender, et al.
Journal of the American Chemical Society, 128, 14814-14815 (2006)
Alexander M Kluwer et al.
Journal of the American Chemical Society, 127(44), 15470-15480 (2005-11-03)
The kinetics of the stereoselective semi-hydrogenation of 4-octyne in THF by the highly active catalyst [Pd{(m,m'-(CF(3))(2)C(6)H(3))-bian}(ma)] (2) (bian = bis(imino)acenaphthene; ma = maleic anhydride) has been investigated. The rate law under hydrogen-rich conditions is described by r = k[4-octyne](0.65)[Pd][H(2)], showing
Chengxiang Zhou et al.
The Journal of organic chemistry, 71(8), 3184-3191 (2006-04-08)
The Pd(II)-catalyzed reaction of arylboronic acids and internal alkynes provides a convenient route to a wide variety of tetrasubstituted olefins. The reaction is conducted in DMSO using molecular O2 as an oxidant in the absence of any base. The reaction
Kelly E Aldrich et al.
Dalton transactions (Cambridge, England : 2003), 48(30), 11352-11360 (2019-07-10)
Highly dehydrated silica gel, SiO2700, gave a material with a total surface hydroxyl density of 0.31 ± 0.05 mmol g-1, 0.9 ± 0.1 Si-OH sites per nm2. Treatment of this material with Ti(NMe2)4 gave Ti(NMe2)3/SiO2700, which is 1.50% ± 0.07
Davide Albani et al.
Nature communications, 9(1), 2634-2634 (2018-07-08)
Ensemble control has been intensively pursued for decades to identify sustainable alternatives to the Lindlar catalyst (PdPb/CaCO3) applied for the partial hydrogenation of alkynes in industrial organic synthesis. Although the geometric and electronic requirements are known, a literature survey illustrates

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service