242500
2-Bromo-1,1-dimethoxyethane
97%
Synonym(s):
Bromoacetaldehyde dimethyl acetal
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
BrCH2CH(OCH3)2
CAS Number:
Molecular Weight:
169.02
Beilstein:
1280132
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
contains
0.2% potassium carbonate as stabilizer
refractive index
n20/D 1.445 (lit.)
bp
148-150 °C (lit.)
density
1.43 g/mL at 25 °C (lit.)
functional group
acetal
bromo
ether
storage temp.
2-8°C
SMILES string
COC(CBr)OC
InChI
1S/C4H9BrO2/c1-6-4(3-5)7-2/h4H,3H2,1-2H3
InChI key
FUSFWUFSEJXMRQ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
2-Bromo-1,1-dimethoxyethane (Bromoacetaldehyde dimethyl acetal) was used in the synthesis of 2,3-O-acetal via reaction with 2,3-diol in the presence of 10-camphorsulphonic acid (CSA).
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
127.4 °F
Flash Point(C)
53 °C
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
1→2 Migration and concurrent glycosidation of phenyl 1-thio-α-mannopyranosides via 2, 3-O-cyclic dioxonium intermediates.
Tetrahedron, 59(2), 249-254 (2003)
Ping Xu et al.
Polymers, 11(2) (2019-04-10)
Hyperbranched polycaprolactone with controlled structure was synthesized by reversible addition-fragmentation chain transfer radical ring-opening polymerization along with self-condensed vinyl polymerization (SCVP) of 2-methylene-1,3-dioxepane (MDO). Vinyl 2-[(ethoxycarbonothioyl) sulfanyl] propanoate (ECTVP) was used as polymerizable chain transfer agent. Living polymerization behavior was
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service