Skip to Content
Merck
All Photos(1)

Key Documents

236314

Sigma-Aldrich

p-Toluidine

99.6%

Synonym(s):

4-Aminotoluene, 4-Methylaniline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4NH2
CAS Number:
Molecular Weight:
107.15
Beilstein:
471281
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
bp:
200 °C (lit.)
vapor pressure:
0.26 mmHg ( 25 °C)
0.66 mmHg ( 38 °C)

vapor density

3.9 (vs air)

vapor pressure

0.26 mmHg ( 25 °C)
0.66 mmHg ( 38 °C)

Assay

99.6%

form

crystalline

autoignition temp.

899 °F

expl. lim.

6.6 %

pH

7.8 (7 g/L)

bp

200 °C (lit.)

mp

41-46 °C (lit.)

solubility

water: soluble 135 part(lit.)
acetone: freely soluble(lit.)
alcohol: freely soluble(lit.)
carbon disulfide: freely soluble(lit.)
diethyl ether: freely soluble(lit.)
methanol: freely soluble(lit.)
oil: freely soluble(lit.)

density

0.973 g/mL at 25 °C (lit.)

SMILES string

Cc1ccc(N)cc1

InChI

1S/C7H9N/c1-6-2-4-7(8)5-3-6/h2-5H,8H2,1H3

InChI key

RZXMPPFPUUCRFN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

p-Toluidine undergoes oxidative coupling reaction with catecholamine to form an orange water-soluble dye during flow injection-spectrophotometric determination of catecholamine drugs.

Application

p-Toluidine has been used in preparation of:
  • aromatic azo compounds
  • bidentate Schiff base ligands via condensation with salicylaldehyde

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Carc. 2 - Eye Irrit. 2 - Skin Sens. 1A

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Flow injection-spectrophotometeric determination of some catecholamine drugs in pharmaceutical preparations via oxidative coupling reaction with< i> p</i>-toluidine and sodium periodate.
Abdulrahman LK, et al.
Analytica Chimica Acta, 538(1), 331-335 (2005)
Ruthenium (II) complexes containing bidentate Schiff bases and their antifungal activity.
Dharmaraj N, et al.
Transition Met. Chem. (London), 26(1-2), 105-109 (2001)
D R Doerge et al.
Chemical research in toxicology, 4(5), 556-560 (1991-09-01)
The metabolism of three arylamine substrates by H2O2 in the presence of each of the peroxidative enzymes chloroperoxidase (CPX) and pea seed peroxygenase (PSM) was conducted with normal H2O2 and with 18O-labeled H2O2. The resulting C-nitroso aromatic metabolites were examined
Abdessamad Grirrane et al.
Nature protocols, 5(3), 429-438 (2010-03-06)
This protocol describes the aerobic oxidation of aromatic anilines to aromatic azo compounds using gold (Au) nanoparticles supported on TiO(2) as a catalyst. Yields above 98% are achieved under a few bars of oxygen pressure. It should be noted that
J C Kennelly et al.
Mutation research, 137(1), 39-45 (1984-07-01)
The conventional Ames assay metabolising system was confirmed to be deficient in its ability to N-acetylate. This may render the test less sensitive to compounds which normally have an acetylation step during their in vivo activation to carcinogens. The addition

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service