Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
p
-Toluidine
Synonym(s):
4-Aminotoluene, 4-Methylaniline,
p
-Toluidine
Linear Formula:
CH
3
C
6
H
4
NH
2
CAS Number:
106-49-0
Molecular Weight:
107.15
EC Number:
203-403-1
Products
Product Category
building blocks (2)
Brand
Sigma-Aldrich (1)
Supelco (2)
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Puriss (1)
Puriss P.A. (1)
pharmaceutical primary standard (1)
Technique
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
236314
p
-Toluidine,
99.6%
Expand
View Pricing
461121
p
-Toluidine,
99%
Expand
View Pricing
1.10841
p
-Toluidine,
GR for analysis
Expand
View Pricing
1672053
p
-Toluidine,
United States Pharmacopeia (USP) Reference Standard
Expand
View Pricing
89630
p
-Toluidine,
for spectrophotometric det. of Au, Tl(III), W, ≥99.0%
Expand
View Pricing