Skip to Content
Merck
All Photos(1)

Key Documents

71320

Sigma-Aldrich

Sodium borohydride

purum p.a., ≥96% (gas-volumetric)

Synonym(s):

Sodium tetrahydridoborate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaBH4
CAS Number:
Molecular Weight:
37.83
EC Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

grade

purum p.a.

Quality Level

Assay

≥96% (gas-volumetric)

form

powder

reaction suitability

reagent type: reductant

mp

>300 °C (dec.) (lit.)

anion traces

chloride (Cl-): ≤5000 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Ca: ≤50 mg/kg
Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
K: ≤1000 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

SMILES string

[BH4-].[Na+]

InChI

1S/BH4.Na/h1H4;/q-1;+1

InChI key

YOQDYZUWIQVZSF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sodium borohydride is considered as a selective reducing agent due to its ability to selectively reduce aldehydes and ketones into their corresponding alcohols in the presence of other functional groups such as nitro, ester, amide, lactone, epoxide, and nitriles that are otherwise susceptible to reduction.[1] It is being considered as a promising candidate for hydrogen storage applications.[2]

Application

Sodium borohydride may be used as a reductant and hydrogen source for the semi-hydrogenation of alkynes to the corresponding alkenes in the presence of nickel nanoparticles generated in situ from nickel chloride and methanol.[3] It can also be used as a reducing agent for the synthesis of gold nanoparticles from gold chloride.[4]

Other Notes

Reduction of carboxylic acid derivatives [5][6]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Repr. 1B - Skin Corr. 1B - Water-react 1

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G.W. Gribble
ACS Symp. Ser., 641, 167-167 (1996)
hemoselective reductions with sodium borohydride.
Ward D
Canadian Journal of Chemistry, 67(7), 1206-1211 (1989)
Synthesis of thiol-derivatised gold nanoparticles in a two-phase liquid?liquid system.
Brust M
Journal of the Chemical Society. Chemical Communications, 7, 801-802 (1994)
Ligand-free nickel-catalyzed semihydrogenation of alkynes with sodium borohydride: a highly efficient and selective process for cis-alkenes under ambient conditions.
Wen X
Chemical Communications (Cambridge, England), 53(39), 5372-5375 (2017)
Thermo-economic analysis of a hydrogen production system by sodium borohydride (NaBH 4).
Rivarolo M
International Journal of Hydrogen Energy, 43(3), 1606-1614 (2018)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service