Skip to Content
Merck
All Photos(2)

Key Documents

235202

Sigma-Aldrich

4-Bromomethyl-7-methoxycoumarin

97%

Synonym(s):

BMC, Br-Mmc

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H9BrO3
CAS Number:
Molecular Weight:
269.09
Beilstein:
187211
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

213-215 °C (lit.)

solubility

chloroform: soluble 20 mg/mL, clear, colorless to faintly yellow

fluorescence

λex 322 nm; λem 395 nm (after derivatization)

SMILES string

COc1ccc2C(CBr)=CC(=O)Oc2c1

InChI

1S/C11H9BrO3/c1-14-8-2-3-9-7(6-12)4-11(13)15-10(9)5-8/h2-5H,6H2,1H3

InChI key

CTENSLORRMFPDH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Bromomethyl-7-methoxycoumarin has been used:
  • as derivatization reagent in determination of 5-fluorouracil (5-FU) in human plasma by HPLC-tandem mass spectrometry
  • as derivatization reagent in fluorescence detection of sodium monofluoroacetate (MFA-Na) in biological samples by HPLC
  • in the synthesis of new caged derivatives of hydrolysis-resistant 8-bromoadenosine cyclic 3′,5′-monophosphate (8-Br-cAMP) and 8-bromoguanosine cyclic 3′,5′-monophosphate (8-Br-cGMP)
  • in TLC and HPLC separation and fluorometric analysis of a wide range of naturally occurring acids, including bile and thromboxane B2
Fluorescent label for fatty acids.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

V Hagen et al.
Journal of photochemistry and photobiology. B, Biology, 53(1-3), 91-102 (2000-02-15)
New caged derivatives of hydrolysis-resistant 8-bromoadenosine cyclic 3',5'-monophosphate (8-Br-cAMP) and 8-bromoguanosine cyclic 3',5'-monophosphate (8-Br-cGMP) are described. The compounds are the axial and equatorial isomers of the (7-methoxycoumarin-4-yl)methyl (MCM) esters of cyclic nucleotides. Synthesis is accomplished by treatment of 4-bromomethyl-7-methoxycoumarin with
Brisa S Fernandes et al.
BMC medicine, 13, 289-289 (2015-12-02)
The neurotrophic hypothesis postulates that mood disorders such as bipolar disorder (BD) are associated with a lower expression of brain-derived neurotrophic factor (BDNF). However, its role in peripheral blood as a biomarker of disease activity and of stage for BD
Analytical Letters, 26, 429-429 (1993)
John E French et al.
Environmental health perspectives, 123(3), 237-245 (2014-11-07)
Inhalation of benzene at levels below the current exposure limit values leads to hematotoxicity in occupationally exposed workers. We sought to evaluate Diversity Outbred (DO) mice as a tool for exposure threshold assessment and to identify genetic factors that influence
J H Wolf et al.
Journal of pharmaceutical and biomedical analysis, 10(2-3), 99-107 (1992-02-01)
A major part of modern analytical problem solving deals with the trace level determination of organic compounds and contaminants in biomedical, food and environmental samples. In the analysis of these samples chromatographic techniques play a predominant role. Unfortunately, however, even

Articles

Nitric oxide (NO) as a signal transporter in neurons, endothelial cells and in the immune system.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service