Skip to Content
Merck
All Photos(1)

Key Documents

203580

Sigma-Aldrich

Lead(II) nitrate

99.999% trace metals basis

Synonym(s):

Lead dinitrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pb(NO3)2
CAS Number:
Molecular Weight:
331.21
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.999% trace metals basis
form:
crystals and lumps
solid

Quality Level

Assay

99.999% trace metals basis

form

crystals and lumps
solid

impurities

≤15.0 ppm Trace Metal Analysis

mp

470 °C (dec.) (lit.)

SMILES string

[PbH2++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/2NO3.Pb/c2*2-1(3)4;/q2*-1;+2

InChI key

RLJMLMKIBZAXJO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Lead(II) nitrate (Pb(NO3)2) is a divalent heavy metal nitrate which can be used in the fabrication of perovskite quantum dot sensitized solar cells (QDSCs). It can also be used in the formation of chemical sensors.
Several new polymeric complexes of lead with dialkylamino derivatives of squaric acid were produced to study their coordination modes as possible prototypes for studies of lanthanide complexes.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1 - Repr. 1A - Skin Sens. 1B - STOT RE 1

Target Organs

Blood,Central nervous system,Immune system,Kidney

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Timothy J Aikin et al.
eLife, 9 (2020-09-18)
A large fraction of human cancers contain genetic alterations within the Mitogen Activated Protein Kinase (MAPK) signaling network that promote unpredictable phenotypes. Previous studies have shown that the temporal patterns of MAPK activity (i.e. signaling dynamics) differentially regulate cell behavior.
Julian Wachstein et al.
PloS one, 7(12), e51747-e51747 (2013-01-10)
Human CD4(+)CD25(+)FoxP3(+) T regulatory cells (Tregs) control effector T cells and play a central role in peripheral tolerance and immune homeostasis. Heat shock protein 70 (HSP70) is a major immunomodulatory molecule, but its effect on the functions of Tregs is
Electrokinetic properties of kaolinite in mono-and multivalent electrolyte solutions
Alkan M, et al.
Microporous and Mesoporous Materials : The Official Journal of the International Zeolite Association, 83(1-3), 51-59 (2005)
Polyaniline nanofiber composites with metal salts: chemical sensors for hydrogen sulfide
Virji S, et al.
Small, 1(6), 624-627 (2005)
Xia Yuan et al.
Theranostics, 10(10), 4589-4605 (2020-04-16)
Rationale: The adverse health effects of nano-particulate pollutants have attracted much attention in recent years. Carbon nanomaterials are recognized as risk factors for prolonged inflammatory responses and diffuse alveolar injury. Previous research indicated a central role of alveolar macrophages in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service