Skip to Content
Merck
All Photos(3)

Key Documents

211176

Sigma-Aldrich

Selenous acid

98%

Synonym(s):

Selenious acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2SeO3
CAS Number:
Molecular Weight:
128.97
EC Number:
MDL number:
UNSPSC Code:
12352301
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

70  °C ((lit.))
70 °C (dec.) (lit.)

solubility

water: 50 mg/mL

density

3.004  g/mL at 25 °C ((lit.))
3.004 g/mL at 25 °C (lit.)

SMILES string

O[Se](O)=O
O[Se](O)=O

InChI

1S/H2O3Se/c1-4(2)3/h(H2,1,2,3)
1S/H2O3Se/c1-4(2)3/h(H2,1,2,3)

InChI key

MCAHWIHFGHIESP-UHFFFAOYSA-N
MCAHWIHFGHIESP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Selenous acid can be used as a reagent to synthesize:
  • α-Ketoacectals by reacting with acetophenones and triethylorthoformate in the presence of boron trifluoride etherate as a catalyst.
  • 4-Formyl-7-methylcoumarin using 4,7-dimethylcoumarin in the presence of xylene as a solvent.
  • Selenium substituted quinoxaline derivatives from 2,3-dichloro quionoxaline.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Seidel et al.
Journal of clinical chemistry and clinical biochemistry. Zeitschrift fur klinische Chemie und klinische Biochemie, 16(7), 407-411 (1978-07-01)
A simple one-vial-method was developed for the quantitative determination of sphingomyelinase activity in human leukocytes and urine, using [14C-methyl] sphingomyelin. The measured activities of healthy control persons show a higher scatter in (n=50) urine (1.2 +/- 0.5 nmol/h . ml
One-pot synthesis of alpha-ketoacetals from aryl methyl ketones in the presence of selenous acid catalyzed by boron trifluoride etherate
Kharkongor I, et al.
Tetrahedron Letters, 56, 4359-4362 (2015)
The synthesis of novel coumarin dyes and the study of their photoreaction properties
Wang T, et al.
Dyes and Pigments, 75, 104-110 (2007)
Ropp RC et al.
Encyclopedia of the Alkaline Earth Compounds null
Thomas G Back et al.
The Journal of organic chemistry, 70(23), 9230-9236 (2005-11-05)
[Reaction: see text]. Several novel organoselenium and tellurium compounds were prepared and evaluated as mimetics of the selenoenzyme glutathione peroxidase, which protects cells from oxidative stress by reducing harmful peroxides with the thiol glutathione. The compounds were tested for catalytic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service